Skip to Content
Merck
All Photos(2)

Key Documents

434264

Sigma-Aldrich

Polyethylene

Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

mol wt

(Ultra-high molecular weight)

avg. part. size

125 μm

mp

146 °C

transition temp

Tm (DSC) 146 °C (at peak)

density

0.94 g/mL at 25 °C

SMILES string

C=C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Additive to unsaturated polyesters, epoxides and other polymers to impart the unique properties of UHMWPE. Used in industrial parts, coatings and wear surfaces at 10-40 wt. %.

Features and Benefits

Polar surface groups assist chemical and physical bonding to matrix material. Contributes superior abrasion resistance, reduced coefficient of friction, enhanced tear strength and improved moisture resistance. Easier to process in particle form than molten UHMWPE.

Physical form

Particle surface is oxidized to contain acid and hydroxy groups.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahtab Memarpour et al.
The journal of adhesive dentistry, 15(1), 85-91 (2013-03-28)
To evaluate the clinical performance of polyethylene fiber for anterior primary teeth restorations in children after 2.5 years, according to modified Ryge criteria. Fifty-five maxillary incisors with extensive caries in 24 Iranian patients (mean age 4.2 years) were restored. All
Lisa K McCarthy et al.
Pediatrics, 132(1), e135-e141 (2013-06-19)
Hypothermia on admission to the NICU is associated with increased mortality in preterm infants. Many newborns are hypothermic on admission despite using polyethylene bags (PBs). Using exothermic mattresses (EMs) in addition to PBs may reduce hypothermia but increase hyperthermia. We
[Technologies for manufacturing ultra-high molecular polyethylene-based porous structures for bone implants].
A V Maksimkin et al.
Meditsinskaia tekhnika, (2)(2), 16-20 (2013-06-26)
Morteza Meftah et al.
The Journal of bone and joint surgery. American volume, 95(13), 1193-1197 (2013-07-05)
Ceramic femoral heads produce less wear of the opposing polyethylene than do metal femoral heads in wear simulation studies. This is a matched-pair analysis of the wear of ceramic and metal femoral heads on conventional polyethylene in uncemented total hip
Thibault Honegger et al.
Lab on a chip, 13(8), 1538-1545 (2013-02-23)
In this paper, we introduce a dielectrophoresis (DEP)-based handling method that allows fine 3D manipulation of beads in suspension using a lab on a chip device. The device consists of two layers of linear electrodes on the top and bottom

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service