Skip to Content
Merck
All Photos(2)

Key Documents

428078

Sigma-Aldrich

Polyethylene

Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

gloss

50 %  (45°C, ASTM D 523)

form

pellets

melt index

1.0 g/10 min (190°C/2.16kg)

mp

100-125 °C

density

0.918 g/mL at 25 °C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Film applications having good drawdown and toughness.

Features and Benefits

No-slip or anti-block additive. Films have good puncture resistance and heat seal strength.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morteza Meftah et al.
The Journal of bone and joint surgery. American volume, 95(13), 1193-1197 (2013-07-05)
Ceramic femoral heads produce less wear of the opposing polyethylene than do metal femoral heads in wear simulation studies. This is a matched-pair analysis of the wear of ceramic and metal femoral heads on conventional polyethylene in uncemented total hip
Alicia E Leadford et al.
Pediatrics, 132(1), e128-e134 (2013-06-05)
Hypothermia contributes to neonatal mortality and morbidity, especially in preterm and low birth weight infants in developing countries. Plastic bags covering the trunk and extremities of very low birth weight infants reduces hypothermia. This technique has not been studied in
Maria C S Inacio et al.
Acta orthopaedica, 84(2), 145-152 (2013-03-15)
There is no substantial clinical evidence for the superiority of alternative bearings in total knee arthroplasty (TKA). We compared the short-term revision risk in alternative surface bearing knees (oxidized zirconium (OZ) femoral implants or highly crosslinked polyethylene (HXLPE) inserts) with
Ingvild Bostadløkken et al.
Ugeskrift for laeger, 175(13), 891-892 (2013-04-16)
We present a case of a 66-year-old woman who had a total knee arthroplasty. Shortly after surgery a 90° rotation of her polyethylene liner took place. The complication was undiagnosed for 1.5 years. To know this problem and be able
Mahtab Memarpour et al.
The journal of adhesive dentistry, 15(1), 85-91 (2013-03-28)
To evaluate the clinical performance of polyethylene fiber for anterior primary teeth restorations in children after 2.5 years, according to modified Ryge criteria. Fifty-five maxillary incisors with extensive caries in 24 Iranian patients (mean age 4.2 years) were restored. All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service