Skip to Content
Merck
All Photos(1)

Documents

430609

Sigma-Aldrich

Lanthanum(III) trifluoromethanesulfonate

99.999% trace metals basis

Synonym(s):

La(OTf)3, Lanthanum tris(trifluoromethanesulfonate), Tris(trifluoromethanesulfonato)lanthanum, Lanthanum(III) triflate, Trifluoromethanesulfonic acid lanthanum salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3La
CAS Number:
Molecular Weight:
586.11
Beilstein:
9009103
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.999% trace metals basis

reaction suitability

core: lanthanum
reagent type: catalyst

SMILES string

[La+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.La/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

WGJJZRVGLPOKQT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

A water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jie Wu et al.
Bioresource technology, 324, 124664-124664 (2021-01-18)
Previous work has shown that sulfonation and oxidation of chemi-thermomechanical pulps (CTMPs) significantly enhanced enzyme accessibility to cellulose while recovering the majority of carbohydrates in the water-insoluble component. In the work reported here, modified (sulfonated and oxidized) CTMPs derived from
The Journal of Organic Chemistry, 59, 3590-3590 (1994)
Dennis M Whitfield
Carbohydrate research, 356, 180-190 (2012-04-25)
The Transition State (TS) for any chemical glycosylation reaction is not known with certainty. Both experimental and computational approaches have been limited due to the complexity of the problem. This work describes a preliminary computational ionization approach using density functional
B V Subba Reddy et al.
Organic letters, 15(3), 464-467 (2013-01-18)
A novel method has been devised for the synthesis of 2,4,5-trisubstituted pyrrole derivatives through the coupling of α-diazoketones with β-enaminoketones and esters using 10 mol % of Cu(OTf)(2). A wide range of 2,3-disubstituted indole derivatives were also prepared from α-diazoketones
Fumiya Urabe et al.
The Journal of organic chemistry, 78(8), 3847-3857 (2013-04-13)
The stereocontrolled total synthesis of (-)-cinatrin C1, a phospholipase A2 inhibitor, has been accomplished. The key feature includes the stereoselective construction of the highly substituted tetrahydrofuran core by In(OTf)3-catalyzed Conia-ene reaction of the oxygen-tethered acetylenic malonic ester followed by dihydroxylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service