Skip to Content
Merck
All Photos(2)

Key Documents

418218

Sigma-Aldrich

Scandium(III) triflate

99%

Synonym(s):

Sc(OTf)3, Scandium(III) trifluoromethanesulfonate, Trifluoromethanesulfonic acid scandium(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sc(SO3CF3)3
CAS Number:
Molecular Weight:
492.16
Beilstein:
8510151
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

powder

reaction suitability

core: scandium
reagent type: catalyst

SMILES string

[Sc+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Sc/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

HZXJVDYQRYYYOR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Scandium(III) triflate is an extremely active, efficient, recoverable and reusable acylation catalyst.[1] Its an important catalyst for the Friedel-Crafts acylation, Diels-Alder reactions and other carbon-carbon bond-forming reactions.[2] It also stereochemically catalyzes the radical polymerization of acrylates.[3] Scandium(III) triflate complex of (4′S,5′S)-2,6-bis[4′-(triisopropylsilyl)oxymethyl-5′-phenyl-1′,3′-oxazolin-2′-yl]pyridine has been employed as catalyst for the asymmetric Friedel-Crafts reaction between substituted indoles and methyl (E)-2-oxo-4-aryl-3-butenoates.[4]

Application

Scandium(III) triflate was used as a catalyst in:
  • Hydrothiolation reaction of aromatic and aliphatic thiols.[5]
  • Selective two-electron reduction of O2 by ferrocene derivatives.[6]
  • Vinylogous Friedel-Crafts alkylation of indoles and pyrroles in water.[7]
  • Synthesis of β-cyanoketones.[8]
  • Combination with triethylsilane to reductively open functionalized pyranoside rings.[9]
  • The key steps of synthesis of bullvalone via a stabilized sulfur ylide.[10]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saya Kakuda et al.
Journal of the American Chemical Society, 137(9), 3330-3337 (2015-02-11)
Mononuclear copper complexes, [(tmpa)Cu(II)(CH3CN)](ClO4)2 (1, tmpa = tris(2-pyridylmethyl)amine) and [(BzQ)Cu(II)(H2O)2](ClO4)2 (2, BzQ = bis(2-quinolinylmethyl)benzylamine)], act as efficient catalysts for the selective two-electron reduction of O2 by ferrocene derivatives in the presence of scandium triflate (Sc(OTf)3) in acetone, whereas 1 catalyzes
Alex R Lippert et al.
Journal of the American Chemical Society, 128(46), 14738-14739 (2006-11-16)
Oligosubstituted bullvalones were synthesized in eight steps from 2,6-cycloheptadienone via a unique Lewis acid catalyzed intramolecular cyclopropanation of a stabilized sulfur ylide, leading directly to the tetracyclic cage structure. Upon exposure to base, the substituted bullvalones tautomerized to a hydroxybullvalene
Okamoto, Y., et al.
Macromolecular Symposia, 183, 83-83 (2002)
Mild reductive opening of aryl pyranosides promoted by scandium(III) triflate.
Hua-Li Qin et al.
Journal of the American Chemical Society, 129(1), 38-39 (2007-01-04)
Giovanni Desimoni et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(12), 3630-3636 (2008-02-29)
The asymmetric Friedel-Crafts reaction between a series of substituted indoles 2 a-l and methyl (E)-2-oxo-4-aryl-3-butenoates 3 a-c has been efficiently catalyzed by the scandium(III) triflate complex of (4'S,5'S)-2,6-bis[4'-(triisopropylsilyl)oxymethyl-5'-phenyl-1',3'-oxazolin-2'-yl]pyridine (pybox; 1). Substituted 4-(indol-3-yl)-2-oxo-4-arylbutyric acid methyl esters 4 a-n were usually formed

Articles

Friedel-Crafts acylation with Lewis acid catalysts forms monoacylated products via electrophilic aromatic substitution of arenes.

Friedel-Crafts acylation with Lewis acid catalysts forms monoacylated products via electrophilic aromatic substitution of arenes.

Friedel-Crafts acylation with Lewis acid catalysts forms monoacylated products via electrophilic aromatic substitution of arenes.

Friedel-Crafts acylation with Lewis acid catalysts forms monoacylated products via electrophilic aromatic substitution of arenes.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service