Skip to Content
Merck
All Photos(3)

Documents

426369

Sigma-Aldrich

TEMPO

purified by sublimation, 99%

Synonym(s):

2,2,6,6-Tetramethylpiperidine 1-oxyl, 2,2,6,6-Tetramethyl-1-piperidinyloxy, free radical, TEMPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18NO
CAS Number:
Molecular Weight:
156.25
Beilstein:
1422418
EC Number:
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

purified by

sublimation

reaction suitability

reagent type: oxidant

mp

36-38 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)CCCC(C)(C)N1[O]

InChI

1S/C9H18NO/c1-8(2)6-5-7-9(3,4)10(8)11/h5-7H2,1-4H3

InChI key

QYTDEUPAUMOIOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For a synthetic protocol using NMP initiators, contributed by Prof. Karen Wooley, please visit our technology spotlight.

Application

Stable nitroxide radical useful in controlling living polymerizations
TEMPO (2,2,6,6-Tetramethyl-1-piperidinyloxy) was used for trapping the styrenyl radical generated from benzoyl peroxide during nitroxide-mediated radical polymerization of styrene. TEMPO-mediated oxidation was employed for the carboxylation of water-resistant nanofibrillated cellulose (NFC) films.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Block copolymer synthesis using a commercially available nitroxide-mediated radical polymerization (NMP) initiator.
Lee NS and Wooley KL.
Material Matters, 5, 8-15 (2010)
Jessica M Hoover et al.
Journal of the American Chemical Society, 135(6), 2357-2367 (2013-01-16)
Homogeneous Cu/TEMPO catalyst systems (TEMPO = 2,2,6,6-tetramethylpiperidine-N-oxyl) have emerged as some of the most versatile and practical catalysts for aerobic alcohol oxidation. Recently, we disclosed a (bpy)Cu(I)/TEMPO/NMI catalyst system (NMI = N-methylimidazole) that exhibits fast rates and high selectivities, even
Akira Isogai et al.
Nanoscale, 3(1), 71-85 (2010-10-20)
Native wood celluloses can be converted to individual nanofibers 3-4 nm wide that are at least several microns in length, i.e. with aspect ratios>100, by TEMPO (2,2,6,6-tetramethylpiperidine-1-oxyl radical)-mediated oxidation and successive mild disintegration in water. Preparation methods and fundamental characteristics
Hayaka Fukuzumi et al.
Carbohydrate polymers, 93(1), 172-177 (2013-03-08)
Various mechanical disintegration conditions in water were applied to 2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO)-oxidized cellulose to prepare TEMPO-oxidized cellulose nanofibrils (TOCNs) of uniform widths ∼4 nm but with three different average lengths, 200, 680, and 1100 nm. The viscosity average degrees of polymerization
Soham Maity et al.
Journal of the American Chemical Society, 135(9), 3355-3358 (2013-02-15)
Nitroolefin is a common and versatile reagent. Its synthesis from olefin is generally limited by the formation of mixture of cis and trans compounds. Here we report that silver nitrite (AgNO2) along with TEMPO can promote the regio- and stereoselective

Articles

Block copolymer synthesis using a commercially available nitroxide-mediated radical polymerization (NMP) initiator

Block copolymer synthesis using a commercially available nitroxide-mediated radical polymerization (NMP) initiator

Block copolymer synthesis using a commercially available nitroxide-mediated radical polymerization (NMP) initiator

Block copolymer synthesis using a commercially available nitroxide-mediated radical polymerization (NMP) initiator

See All

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service