Skip to Content
Merck
All Photos(1)

Key Documents

420832

Sigma-Aldrich

LUDOX® AS-30 colloidal silica

30 wt. % suspension in H2O

Synonym(s):

Silica preparation

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
NA.22

description

colloidal

Quality Level

form

suspension

concentration

30 wt. % suspension in H2O

surface area

~230 m2/g

pH

9.1

density

1.2 g/mL at 25 °C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LUDOX® colloidal silica are aqueous dispersions of silica particles that can improve the product performance as an inorganic binder.

Application

Ammonium stabilizing counterion

Legal Information

LUDOX is a registered trademark of W.R. Grace & Co.-Conn.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaswanth Kuthati et al.
Journal of nanoscience and nanotechnology, 13(4), 2399-2430 (2013-06-15)
The advent of Nanotechnology has paved a way for improved disease treatment strategies, the most noteworthy being the mesoporous silica nanoparticles (MSNs) which have gained much recent attention in the field of cancer therapy and its diagnosis. The flaws of
Graham J Edgar et al.
Nature, 506(7487), 216-220 (2014-02-07)
In line with global targets agreed under the Convention on Biological Diversity, the number of marine protected areas (MPAs) is increasing rapidly, yet socio-economic benefits generated by MPAs remain difficult to predict and under debate. MPAs often fail to reach
Linda Kachuri et al.
International journal of cancer, 135(1), 138-148 (2013-11-26)
Crystalline silica is a recognized carcinogen, but the association with lung cancer at lower levels of exposure has not been well characterized. This study investigated the relationship between occupational silica exposure and lung cancer and the combined effects of cigarette
Andrea F G Gargano et al.
Journal of chromatography. A, 1351, 70-81 (2014-06-05)
Mucin glycoproteins belong to a class of high molecular weight, heavy glycosylated, proteins that together with water, salts and lipids constitute mucous secretions. Particular disease states (e.g. obstructive chronic bronchitis and ovarian tumor) are known to modify the composition and
Arianne Soliven et al.
Journal of chromatography. A, 1351, 56-60 (2014-06-08)
Analytical scale active flow technology first generation silica monolithic columns kitted out in curtain flow mode of operation were studied for the first time. A series of tests were undertaken assessing the column efficiency, peak asymmetry and detection sensitivity. Two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service