Merck
  • Occupational exposure to crystalline silica and the risk of lung cancer in Canadian men.
Occupational exposure to crystalline silica and the risk of lung cancer in Canadian men.
International journal of cancer (2013-11-26)
Linda Kachuri, Paul J Villeneuve, Marie-Élise Parent, Kenneth C Johnson, Shelley A Harris
ABSTRACT
Crystalline silica is a recognized carcinogen, but the association with lung cancer at lower levels of exposure has not been well characterized. This study investigated the relationship between occupational silica exposure and lung cancer and the combined effects of cigarette smoking and silica exposure on lung cancer risk. A population-based case-control study was conducted in eight Canadian provinces between 1994 and 1997. Self-reported questionnaires were used to obtain a lifetime occupational history and information on other risk factors. Occupational hygienists assigned silica exposures to each job based on concentration, frequency and reliability. Data from 1681 incident lung cancer cases and 2053 controls were analyzed using logistic regression to estimate odds ratios (OR) and their 95% confidence intervals (CI). Models included adjustments for cigarette smoking, lifetime residential second-hand smoke and occupational exposure to diesel and gasoline engine emissions. Relative to the unexposed, increasing duration of silica exposure at any concentration was associated with a significant trend in lung cancer risk (OR ≥ 30 years: 1.67, 1.21-2.24; ptrend  = 0.002). The highest tertile of cumulative silica exposure was associated with lung cancer (OR = 1.81, 1.34-2.42; ptrend  = 0.004) relative to the lowest. Men exposed to silica for ≥30 years with ≥40 cigarette pack-years had the highest risk relative to those unexposed with <10 pack-years (OR = 42.53, 23.54-76.83). The joint relationship with smoking was consistent with a multiplicative model. Our findings suggest that occupational exposure to silica is a risk factor for lung cancer, independently from active and passive smoking, as well as from exposure to other lung carcinogens.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O