Skip to Content
Merck
All Photos(3)

Key Documents

S5130

Sigma-Aldrich

Silica, fumed

powder

Synonym(s):

Silica, Silicic anhydride, Silicon dioxide, Silicon dioxide amorphous

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

Quality Level

surface area

395 m2/g (±25 m2/g)

refractive index

n20/D 1.46 (lit.)

density

2.3 lb/cu.ft at 25 °C (bulk density)(lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fumed silica may be synthesized by high temperature hydrolysis of SiCl4 in O2(N2)/H2 flame. It is amorphous in nature and possesses very high specific area. The micro droplets of amorphous silica fuse into a branch and form a chain like agglomerate.

Application

Fumed silica is used to deplete lipids and hormones in Taqman assays of human hepatoma cells HuH-7 and human liver cell line HepG2.[1] Preparation of epoxidized natural rubber/fumed silica composites have been reported.[2] Interaction of Gemini surfactant C12-s-C122Br with aqueous suspension of fumed silica has been studied.[3] Fumed silica based nanocomposites reinforced with organo clay may be prepared.[4] TiO2/fumed silica porous ceramic material was used to prepare photocatalytic materials.[5]

Features and Benefits

Has interesting thickening and thixotropic properties, and an enormous external surface area.

Other Notes

Particle sizes are for primary particles, which form branched, chain-like aggregates a few tenths of a micron long.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Interaction Between Fumed-Silica and Epoxidized Natural Rubber
Luo YY, et al.
Journal of Inorganic and Organometallic Polymers and Materials, 21(4), 777-783 (2011)
Melt rheology of organoclay and fumed silica nanocomposites
Ph. Cassagnau
Polymer, 49(9), 2183-2196 (2008)
Preparation and properties of TiO2/fumed silica composite photocatalytic materials
Yu Y, et al.
Procedia Engineering, 27, 448-456 (2012)
Petra Thulin et al.
International journal of molecular medicine, 21(6), 819-824 (2008-05-29)
The peroxisome proliferator-activated receptor delta (PPARdelta) is a transcription factor that regulates genes of importance in lipid and glucose metabolism. ApoA-II is one of the major proteins of the HDL-particle. The aim of this study was to investigate the regulation
Zheng O, et al.
Energy Procedia, 16 Part B, 1211-1219 (2012)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service