Skip to Content
Products
Cart
0
AE
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Back
Maltodextrin
Synonym(s):
Dextrin, Low DE Maltodextrin, MLD, Maltodextrin Powder, Starch Hydrosylate
CAS Number:
9050-36-6
EC Number:
232-940-4
Products
Product Category
carbohydrates
Brand
Sigma-Aldrich
Biological Source
Physical Form
Tissue
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
419672
Maltodextrin,
Expand
View Pricing
419699
Maltodextrin,
Expand
View Pricing
419680
Maltodextrin,
Expand
View Pricing