Skip to Content
Merck
All Photos(3)

Key Documents

290068

Sigma-Aldrich

Zinc trifluoromethanesulfonate

greener alternative

98%

Synonym(s):

Bis(trifluoromethanesulfonato)zinc, Zinc trifluoromethylsulfonate, Zn(OTf)2, Trifluoromethanesulfonic acid zinc salt, Zinc triflate, Zn(OTf)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)2Zn
CAS Number:
Molecular Weight:
363.53
Beilstein:
4028195
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

reaction suitability

core: zinc
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

≥300 °C (lit.)

greener alternative category

SMILES string

[Zn++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/2CHF3O3S.Zn/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2

InChI key

CITILBVTAYEWKR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Zinc trifluoromethanesulfonate acts as a lewis acid in nucleophilic addition and as a catalyst in aldol reactions.

Application

  • Initiating a high-temperature zinc ion battery through a triazolium-based ionic liquid: This study explores the use of zinc trifluoromethanesulfonic acid in ionic liquid for high-temperature zinc ion batteries, demonstrating significant improvements in conductivity and stability. (Li et al., 2022).
  • Enhancing the solubility of 6-mercaptopurine by formation of ionic cocrystal with zinc trifluoromethanesulfonate: The research focuses on improving the solubility of 6-mercaptopurine by forming ionic cocrystals with zinc trifluoromethanesulfonate, resulting in single-crystal-to-single-crystal transformation. (Yao et al., 2014).
  • Spontaneous desaturation of the solvation sheath for high-performance anti-freezing zinc-ion gel-electrolyte: This paper discusses the development of a high-performance anti-freezing zinc-ion gel-electrolyte using zinc trifluoromethanesulfonate, which shows excellent performance even at low temperatures. (Li et al., 2023).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zinc triflate as Lewis acid in nucleophilic addition to cyclic N-acyliminium ions
Ronaldo Aloise P et al.
Synlett, 2005, 2297-2300 (2005)
Magnesium and zinc-catalyzed thioketalization
Corey EJ and Shimoji K
Tetrahedron Letters, 24, 169-169 (1983)
Novel heterogeneous zinc triflate catalysts for the rearrangement of ?-pinene oxide.
Wilson K, et al.
Catalysis Letters, 61(1-2), 51-55 (1999)
Regioselective synthesis of 3-alkylindoles mediated by zinc triflate.
Zhu X and Ganesan A
The Journal of Organic Chemistry, 67(8), 2705-2708 (2002)
Gang Liu et al.
Chemical communications (Cambridge, England), 48(56), 7049-7051 (2012-06-12)
An unexpected silver triflate-catalyzed tandem reaction of N'-(2-alkynylbenzylidene)hydrazide with ketene through 6-endo-cyclization, [3+2] cycloaddition and rearrangement is reported. This reaction proceeds efficiently to generate the molecular complexity with the formation of four bonds in a one-pot procedure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service