Skip to Content
Merck
All Photos(3)

Key Documents

407097

Sigma-Aldrich

1-Bromo-2,4,6-tri-tert-butylbenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]3C6H2Br
CAS Number:
Molecular Weight:
325.33
Beilstein:
1913257
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

168-173 °C (lit.)

functional group

bromo

SMILES string

CC(C)(C)c1cc(c(Br)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C18H29Br/c1-16(2,3)12-10-13(17(4,5)6)15(19)14(11-12)18(7,8)9/h10-11H,1-9H3

InChI key

JOKZWHPYNRDCOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-2,4,6-tri-tert-butylbenzene (2,4,6-tri-tert-butylbromobenzene) is a hindered aryl bromide. 1-Bromo-2,4,6-tri-tert-butylbenzene on reaction with phenylboronic acid yields α,α-dimethyl-β-phenyl hydrostyrene.

Application

1-Bromo-2,4,6-tri-tert-butylbenzene was used in the synthesis of bulky biarylphosphine ligand. This ligand was reported to participate in the Pd-catalyzed C-O cross-coupling of a wide range of aryl halides and phenols under milder conditions. It was used to investigate the effect on oligomerization of increased steric bulk in dimethylindium(III) chalcogenolates. It may be used to form α,α-dimethyl-β-phenyl hydrostyrene by reacting with phenylboronic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The first Cu-and amine-free Sonogashira-type cross-coupling in the C-6-alkynylation of protected 2'-deoxyadenosine.
Ngassa FN, et al.
Tetrahedron, 65(21), 4085-4091 (2009)
Luca Salvi et al.
Organic letters, 14(1), 170-173 (2011-12-21)
A new bulky biarylphosphine ligand (L8) has been developed that allows the Pd-catalyzed C-O cross-coupling of a wide range of aryl halides and phenols under milder conditions than previously possible. A direct correlation between the size of the ligand substituents
Glen G Briand et al.
Dalton transactions (Cambridge, England : 2003), 39(16), 3833-3841 (2010-04-08)
The effect on oligomerization of increased steric bulk in dimethylindium(III) chalcogenolates (Me(2)InER') (E = O, S, Se) has been examined. The facile reaction of Me(3)In with a series of phenols, thiophenols and selenophenols afforded the compounds [Me(2)InO(C(6)H(5))](2) (1), [Me(2)InO(2,6-Me(2)C(6)H(3))](2) (2)
Palladium(0)-catalyzed intermolecular amination of unactivated C(sp³)-H bonds.
Jun Pan et al.
Angewandte Chemie (International ed. in English), 50(37), 8647-8651 (2011-08-04)

Articles

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service