Skip to Content
Merck
All Photos(2)

Documents

223778

Sigma-Aldrich

1,3,5-Tri-tert-butylbenzene

97%

Synonym(s):

1,3,5-Tri-t-butylbenzene, 1,3,5-Tris(1,1-dimethylethyl)benzene, 1,3,5-Tris(tert-butyl)benzene, 1,3,5-Tritert-butylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3[C(CH3)3]3
CAS Number:
Molecular Weight:
246.43
Beilstein:
1909579
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

121-122 °C/12 mmHg (lit.)

mp

67-72 °C (lit.)

SMILES string

CC(C)(C)c1cc(cc(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C18H30/c1-16(2,3)13-10-14(17(4,5)6)12-15(11-13)18(7,8)9/h10-12H,1-9H3

InChI key

GUFMBISUSZUUCB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The Sandros-Boltzmann (SB) dependency on the reaction free energy of 1,3,5-tri-tert-butylbenzene has been studied.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Alan Rodrigo García-Cicourel et al.
Journal of chromatography. A, 1590, 113-120 (2019-01-19)
Highly purified mineral oils are used in several pharmaceutical, foods and cosmetics applications. A fast and simple method was developed for the analysis of the total level of residual mineral oil aromatic hydrocarbons (MOAH) in these oils and in the
Eun Chul Pack et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 37(5), 858-868 (2020-03-12)
Mineral oil hydrocarbons (MOH) are widely used in the food industry for applications such as printing inks, additives, adhesives, and processing aids for food additives. Recently, the migration of MOH from food contact paper and board into foods has raised
Toshitaka Funazukuri et al.
Journal of chromatography. A, 1527, 91-96 (2017-11-05)
Retention factors for two enantiomers of trans-stilbene oxide, k
Federica Ianni et al.
Journal of chromatography. A, 1557, 20-27 (2018-05-05)
In the present paper we describe the optimization and the application of a chromatographic method suitable to get all four diastereoisomers of C24-C25 cyclopropyl dafachronic acid derivatives in sufficient amount for their biological appraisal towards the nuclear hormone receptor transcription
Patrick Tarnow et al.
PloS one, 11(1), e0147239-e0147239 (2016-01-16)
The majority of printing inks are based on mineral oils (MOs) which contain complex mixtures of saturated and aromatic hydrocarbons. Consumer exposure to these oils occurs either through direct skin contacts or, more frequently, as a result of MO migration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service