Skip to Content
Merck
All Photos(1)

Key Documents

481408

Sigma-Aldrich

Chlorodicyclohexylphosphine

97%

Synonym(s):

Dicyclohexylchlorophosphine, Dicyclohexylphosphine chloride, Dicyclohexylphosphinous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2PCl
CAS Number:
Molecular Weight:
232.73
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.533 (lit.)

bp

165 °C/12 mmHg (lit.)

density

1.054 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

ClP(C1CCCCC1)C2CCCCC2

InChI

1S/C12H22ClP/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

AKJFBIZAEPTXIL-UHFFFAOYSA-N

Application

Chlorodicyclohexylphosphine can be used as a reactant in the synthesis of:
  • 1,2-Bis(dicyclohexylphosphinoxy)ethane ligand by reacting with ethylene glycol in the presence of triethylamine via Michaelis−Arbuzov type rearrangements.
  • 1,1,2,2-tetracyclohexyldiphosphine monosulfide ligand by treating with LiS.

It can be also used as a starting material for the preparation of some other ligands such as dicyclohexylphosphine oxide , phosphino substituted N-aryl pyrroles , di-tert-butyl((dicyclohexylphosphino)methyl)phosphine , dicyclohexylcyclopentylphosphine. These ligands are used in Pd-catalyzed cross-coupling reactions.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Intramolecular alkyl phosphine dehydrogenation in cationic rhodium complexes of tris (cyclopentylphosphine)
Douglas TM, et al.
Chemistry?A European Journal , 14(3), 1004-1022 (2008)
Latexin deletion protects against radiation-induced hematopoietic damages via selective activation of Bcl-2 prosurvival pathway.
Zhang, et al.
Haematologica (2023)
Synthesis of 1, 2-bis [(diorgano) phosphino] ethanes via Michaelis-Arbuzov type rearrangements
Baldwin LC and Fink MJ
Journal of Organometallic Chemistry, 646(1-2), 230-238 (2002)
Sterically crowded diphosphinomethane ligands: molecular structures, UV-photoelectron spectroscopy and a convenient general synthesis of t Bu 2 PCH 2 P t Bu 2 and related species
Eisentrager, F, et al.
New. J. Chem., 27(3), 540-550 (2003)
A sulfur mimic of 1, 1-bis (diphenylphosphino) methane: a new ligand opens up
Sues PE, et al.
Chemical Communications (Cambridge, England), 50(36), 4707-4710 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service