Skip to Content
Merck
All Photos(3)

Documents

346276

Sigma-Aldrich

Tetrakis(acetonitrile)copper(I) hexafluorophosphate

97%

Synonym(s):

Copper(I) tetrakis(acetonitrile) hexafluorophosphate, Cuprous tetrakis(acetonitrile) hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3CN)4Cu]PF6
CAS Number:
Molecular Weight:
372.72
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: copper
reagent type: catalyst

mp

160 °C (dec.) (lit.)

SMILES string

[Cu+].CC#N.CC#N.CC#N.CC#N.F[P-](F)(F)(F)(F)F

InChI

1S/4C2H3N.Cu.F6P/c4*1-2-3;;1-7(2,3,4,5)6/h4*1H3;;/q;;;;+1;-1

InChI key

GNQXUMGHBSAQBV-UHFFFAOYSA-N

Application

Tetrakis(acetonitrile)copper(I) hexafluorophosphate can be employed as a catalyst for the synthesis of triazolo[1,2-a]indazole-1,3,8-trione and 2H-indazolo[2,1-b]phthalazine-trione derivatives by the reaction of aryl aldehydes, dimedone and urazole or phthalhydrazide, respectively.
It can also be used as a copper precursor:
  • For the formation of Cu2S/CdS nanorod junctions by cationic exchange process.
  • To synthesize monometallic copper macrocyclic biquinazoline ligand [Cu(Mabiq)].
  • To prepare the cuprous complex [Cu(2-iQBO)(POP)]PF6]; where 2-iQBO=2-(1′-isoquinolyl)benzoxazole and POP=bis[2-(diphenylphosphino)phenyl]ether, which can form luminescent pseudo-polymorphs.
  • For the synthesis of [Cu(NHC)2]PF6 complexes [(NHC = N-heterocyclic carbene] which can be further used in the hydrosilylation of carbonyl compounds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural Characterization and Photochemical Properties of Mono-and Bimetallic Cu-Mabiq Complexes.
Stark HS, et al.
Inorganic Chemistry, 28(9), 095705-095705 (2018)
Tetrakis (acetonitrile) copper (I) Hexafluorophosphate as an Efficient Catalyst for the Synthesis of Triazolo [1, 2-a] indazole-1, 3, 8-trione and 2Hindazolo [2, 1-b] phthalazine-trione Derivatives.
Azarifar D, et al.
Letters in Organic Chemistry, 9(2), 128-132 (2012)
Synthesis and characterization of [Cu (NHC)2] X complexes: Catalytic and mechanistic studies of hydrosilylation reactions.
Diez-Gonzalez S, et al.
Chemistry?A European Journal , 14(1), 158-168 (2008)
Two luminescent pseudo-polymorphic cuprous complexes with different optical properties: Synthesis, characterization and TD-DFT calculations.
Tao XD, et al.
Polyhedron, 144(9), 36-43 (2018)
Biswajit Kundu et al.
Nanotechnology, 28(9), 095705-095705 (2017-01-31)
Through scanning tunneling spectroscopy, we envisage imaging a heterostructure, namely a junction formed in a single nanorod. While the differential conductance spectrum provides location of conduction and valence band edges, dI/dV images record energy levels of materials. Such dI/dV images

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service