382140
Diethyl 2,5-dihydroxyterephthalate
97%
Synonym(s):
2,5-Dihydroxyterephthalic acid diethyl ester
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(HO)2C6H2(CO2CH2CH3)2
CAS Number:
Molecular Weight:
254.24
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
97%
mp
135-137 °C (lit.)
SMILES string
CCOC(=O)c1cc(O)c(cc1O)C(=O)OCC
InChI
1S/C12H14O6/c1-3-17-11(15)7-5-10(14)8(6-9(7)13)12(16)18-4-2/h5-6,13-14H,3-4H2,1-2H3
InChI key
UQOUOXLHXPHDHF-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Gerard Boix et al.
ACS applied materials & interfaces, 12(9), 10554-10562 (2020-02-07)
Pollution of water with heavy metals is a global environmental problem whose impact is especially severe in developing countries. Among water-purification methods, adsorption of heavy metals has proven to be simple, versatile, and cost-effective. However, there is still a need
Harley D Betts et al.
Molecules (Basel, Switzerland), 25(19) (2020-10-07)
Silver(I)-based coordination polymers or metal-organic frameworks (MOFs) display useful antibacterial properties, whereby distinct materials with different bonding can afford control over the release of silver(I) ions. Such silver(I) materials are comprised of discrete secondary building units (SBUs), and typically formed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service