Skip to Content
Merck
All Photos(1)

Key Documents

381446

Sigma-Aldrich

Iron(III) oxalate hexahydrate

Synonym(s):

Ferric oxalate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2(C2O4)3 · 6H2O
CAS Number:
Molecular Weight:
483.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks

form

powder and chunks

Quality Level

concentration

22-25% Fe (by Na2S2O3, titration)

application(s)

battery manufacturing

SMILES string

O.O.O.O.O.O.O=C1O[Fe](OC1=O)OC(=O)C(=O)O[Fe]2OC(=O)C(=O)O2

InChI

1S/3C2H2O4.2Fe.6H2O/c3*3-1(4)2(5)6;;;;;;;;/h3*(H,3,4)(H,5,6);;;6*1H2/q;;;2*+3;;;;;;/p-6

InChI key

FWXIZVVTJVNNRX-UHFFFAOYSA-H

Application

Iron(III) oxalate hexahydrate may be used to prepare amorphous Fe2O3 nanoparticles of desired properties and α -Fe2O3 for electrochemical Li ion batteries.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron oxide nanoparticle powders with high surface area
Krizek M, et al.
AIP Conference Proceedings, 1489.1, 88-88 (2012)
Facile fabrication of hierarchical ?-Fe2O3: self-assembly and its magnetic and electrochemical properties.
Muruganandham M, et al.
The Journal of Physical Chemistry C, 115.37, 18164-18173 (2011)

Articles

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service