Skip to Content
Merck
All Photos(1)

Key Documents

268259

Sigma-Aldrich

Nickel

foil, thickness 0.125 mm, ≥99.9%

Synonym(s):

Ni

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥99.9%

form

foil

reaction suitability

core: nickel
reagent type: catalyst

resistivity

6.97 μΩ-cm, 20°C

thickness

0.125 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

Application

Common applications of Nickel in synthetic chemistry:
  • High-quality graphene films can be synthesized on nickel foil by rapid thermal chemical vapor deposition.[1]
  • Nickel foil can be used as a substrate to grow piezoelectric thin films of Pb(Zr,Ti)O3 (PZT).[2]
  • Nickel oxide nanowires can be synthesized by calcining nickel foil after soaking in lithium hydroxide solution.[3]
  • Manganese dioxide can be electrodeposited on a nickel foil for fabricating thin film ultracapacitors.[4]

Quantity

150g ~13 x (100x100mm)
25g ~2 x (100x100mm)

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Synthesis of high-quality graphene films on nickel foils by rapid thermal chemical vapor deposition.
Huang L, et al.
Carbon, 50(2), 551-556 (2012)
Novel electrode materials for thin?film ultracapacitors: comparison of electrochemical properties of sol?gel?derived and electrodeposited manganese dioxide.
Pang SC, et al.
Journal of the Electrochemical Society, 147(2), 444-450 (2000)
{001} Oriented piezoelectric films prepared by chemical solution deposition on Ni foils.
Yeo HG and Trolier-McKinstry S.
Journal of Applied Physics, 116(1), 014105-014105 (2014)
Ornella Valenti et al.
Cerebral cortex (New York, N.Y. : 1991), 19(3), 658-674 (2008-07-18)
The prefrontal cortex receives multiple inputs from the hippocampal complex, which are thought to drive memory-guided behavior. Moreover, dysfunctions of both regions have been repeatedly associated with several psychiatric disorders. Therefore, understanding the interconnections and modulatory interactions between these regions
A J Feilzer et al.
Nederlands tijdschrift voor tandheelkunde, 120(6), 335-341 (2013-07-19)
Orally applied metal alloys can cause undesirable physical effects. A distinction needs to be made in this respect between local and systemic reactions and toxic and immunological reactions. A case is presented which illustrates this problem. In this case, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service