Skip to Content
Merck
All Photos(1)

Key Documents

334537

Sigma-Aldrich

Potassium perruthenate

Synonym(s):

Potassium ruthenium oxide, Potassium ruthenium tetraoxide, Potassium tetraoxoruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KRuO4
CAS Number:
Molecular Weight:
204.17
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid
solubility:
H2O: slightly soluble(lit.)

form

solid

Quality Level

reaction suitability

reagent type: oxidant

concentration

≥49.0% (gravimetric)

solubility

H2O: slightly soluble(lit.)

SMILES string

[K+].[O-][Ru](=O)(=O)=O

InChI

1S/K.4O.Ru/q+1;;;;-1;

InChI key

RDFCRPAXZNYVJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun-Ang Raiber et al.
Genome biology, 13(8), R69-R69 (2012-08-21)
Methylation of cytosine in DNA (5mC) is an important epigenetic mark that is involved in the regulation of genome function. During early embryonic development in mammals, the methylation landscape is dynamically reprogrammed in part through active demethylation. Recent advances have
Chan-Wang J Lio et al.
Science immunology, 4(34) (2019-04-28)
TET enzymes are dioxygenases that promote DNA demethylation by oxidizing the methyl group of 5-methylcytosine to 5-hydroxymethylcytosine (5hmC). Here, we report a close correspondence between 5hmC-marked regions, chromatin accessibility and enhancer activity in B cells, and a strong enrichment for
Jay P Patel et al.
Chemical research in toxicology, 28(12), 2352-2363 (2015-10-09)
The DNA of all organisms is metabolically active due to persistent endogenous DNA damage, repair, and enzyme-mediated base modification pathways important for epigenetic reprogramming and antibody diversity. The free bases released from DNA either spontaneously or by base excision repair
Michael W Cooke et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(26), 14703-14711 (2020-06-17)
The undeclared release and subsequent detection of ruthenium-106 (106Ru) across Europe from late September to early October of 2017 prompted an international effort to ascertain the circumstances of the event. While dispersion modeling, corroborated by ground deposition measurements, has narrowed
Yibin Liu et al.
Nature biotechnology, 37(4), 424-429 (2019-02-26)
Bisulfite sequencing has been the gold standard for mapping DNA modifications including 5-methylcytosine (5mC) and 5-hydroxymethylcytosine (5hmC) for decades1-4. However, this harsh chemical treatment degrades the majority of the DNA and generates sequencing libraries with low complexity2,5,6. Here, we present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service