Skip to Content
Merck
All Photos(6)

Key Documents

569747

Sigma-Aldrich

Grubbs Catalyst® M204

Umicore

Synonym(s):

(1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene)dichloro(phenylmethylene)(tricyclohexylphosphine)ruthenium, Benzylidene[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(tricyclohexylphosphine)ruthenium, Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene](benzylidene)(tricyclohexylphosphine)ruthenium(II), Grubbs Catalyst® 2nd Generation, Grubbs Catalyst® M2a (C848)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H65Cl2N2PRu
CAS Number:
Molecular Weight:
848.97
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Ring-Opening Polymerization

mp

143.5-148.5 °C

storage temp.

2-8°C

SMILES string

CC1=CC(C)=CC(C)=C1N2CCN(C3=C(C)C=C(C)C=C3C)C2=[Ru](Cl)(Cl)=CC4=CC=CC=C4.P(C5CCCCC5)(C6CCCCC6)C7CCCCC7

InChI

1S/C21H26N2.C18H33P.C7H6.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-7-5-3-2-4-6-7;;;/h9-12H,7-8H2,1-6H3;16-18H,1-15H2;1-6H;2*1H;/q;;;;;+2/p-2

InChI key

FCDPQMAOJARMTG-UHFFFAOYSA-L

Application

Grubbs Catalyst® M204 can be used as a catalyst for ring-closing metathesis (RCM), cross-metathesis, and ring-opening metathesis polymerization (ROMP). It is also used to synthesize trisubstituted olefins with excellent functional group tolerance and selectivity via cross-metathesis and ring closing metathesis reactions.
It can also be used as a catalyst:

  • To synthesize coumarins from phenolic compounds via RCM.
  • To cleave secondary (E)-allyl vic-diols to aldehydes.
For small scale and high throughput uses, product is also available as ChemBeads (919764)

Learn more about our metathesis catalysts

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly selective cross-metathesis with phenyl vinyl sulphone using the `second generation?Grubbs? catalyst
Grela K and Bieniek M
Tetrahedron Letters, 42(36), 6425-6428 (2001)
D L Wright et al.
Organic letters, 3(26), 4275-4277 (2002-01-11)
The ring-opening cross-metathesis of oxabicyclo[3.2.1]octene derivatives provides a convenient method for preparing differentially substituted 4-pyrones. The major competing reaction is the ring-opening metathesis polymerization of the bridged olefin. Studies on this reaction have shown that substituents on the bicyclic alkene
A modified three-field technique for breast treatment.
Svensson GK, et al.
International Journal of Radiation Oncology, Biology, Physics, 6(6), 689-694 (1980)
Novel cleavage of (E)-allyl vic-diols to aldehydes using the 2nd-generation Grubbs catalyst
H Chunguang and U Daisuke
Tetrahedron Letters, 49(49), 6988-6990 (2008)
Total synthesis of amphidinolide Y by formation of trisubstituted (E)-double bond via ring-closing metathesis of densely functionalized alkenes
Jin Jian, et al.
Organic Letters, 9(13), 2585-2588 (2007)

Articles

Learn tips and tricks for how to properly use inhibitors including how to select the right inhibitor and how to plan experiments with inhibitors.

Learn tips and tricks for how to properly use inhibitors including how to select the right inhibitor and how to plan experiments with inhibitors.

Learn tips and tricks for how to properly use inhibitors including how to select the right inhibitor and how to plan experiments with inhibitors.

Learn tips and tricks for how to properly use inhibitors including how to select the right inhibitor and how to plan experiments with inhibitors.

Protocols

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

Related Content

Research focuses on ruthenium alkylidene complexes for versatile olefin metatheses catalysis in the Grubbs group.

Research focuses on ruthenium alkylidene complexes for versatile olefin metatheses catalysis in the Grubbs group.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service