Skip to Content
Merck
All Photos(1)

Key Documents

32598

Sigma-Aldrich

Diallyldimethylammonium chloride

≥97.0% (AT)

Synonym(s):

DADMAC, Dimethyldiallylammonium chloride, N,N-Diallyl-N,N-dimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2=CHCH2)2N(Cl)(CH3)2
CAS Number:
Molecular Weight:
161.67
Beilstein:
5830817
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥97.0% (AT)

form

solid

mp

140-148 °C

SMILES string

[Cl-].C[N+](C)(CC=C)CC=C

InChI

1S/C8H16N.ClH/c1-5-7-9(3,4)8-6-2;/h5-6H,1-2,7-8H2,3-4H3;1H/q+1;/p-1

InChI key

GQOKIYDTHHZSCJ-UHFFFAOYSA-M

General description

Diallyldimethylammonium chloride (DADMAC) is a quaternary ammonium compound available in solid form with an assay of ≥97.0%. It serves as a versatile monomer in various polymerization processes and is primarily used in the production of cationic polymers. These polymers are essential in a range of applications, particularly in the fields of water treatment, paper production, and personal care products. Additionally, DADMAC exhibits antifouling properties, making it valuable for applications where the prevention of biofilm formation and surface fouling is critical.[1][2][3]

Application

Diallyldimethylammonium chloride can be used as:
  • A precursor for the synthesis of poly(diallyldimethylammonium chloride) (PDADMAC), which is widely used in various applications related to water purification and wastewater management.
  • a monomer in the synthesis of cationic hydrogels via radical polymerization. The resulting polymer, poly(diallyldimethylammonium chloride) (PDADMAC), features a high cationic charge density, which is crucial for effectively binding anionic dyes. This property makes PDADMAC-based hydrogels valuable materials for wastewater treatment applications.
  • DADMAC has been used in dental biomaterials to enhance their antifouling properties. When incorporated into acrylic resins, it provides effective antibacterial and antifungal activity without compromising biocompatibility

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanyuan Zhang et al.
ACS nano, 9(7), 7124-7132 (2015-07-15)
In this study, we demonstrate multilevel and multicomponent layer-by-layer (LbL) assembly as a convenient and generally applicable method for the fabrication of nanofibrillar films by exploiting the dynamic nature of polymeric complexes. The alternate deposition of poly(allylamine hydrochloride)-methyl red (PAH-MR)
Deyi Zhu et al.
Journal of industrial microbiology & biotechnology, 42(2), 189-196 (2014-12-30)
Collagen fiber (CF), an abundant natural biopolymer, features many favorable properties that make it a potential carrier for cell immobilization. In the present investigation, CF was grafted with polyethyleneimine (PEI) using glutaraldehyde (GA) as the cross-linking agent, resulting in the
Zhenxi Zhang et al.
Journal of biomedical optics, 20(5), 51043-51043 (2015-05-30)
There are three possible mechanisms for 5-aminolevulinic acid (5-ALA) conjugated gold nanoparticles (GNPs) through electrostatic bonding for photodynamic therapy (PDT) of cancer: GNPs delivery function,singlet oxygen generation (SOG) by GNPs irradiated by light, and surface resonance enhancement (SRE) of SOG.
Ya Xiong et al.
Talanta, 129, 282-289 (2014-08-17)
Simple, rapid and sensitive analysis of thrombin (a tumor biomarker) in complex samples is quite clinical relevant and essential for the development of disease diagnosis and pharmacotherapy. Herein, we developed a novel method based on aptamer-conjugated magnetic graphene/gold nanoparticles nanocomposites
Feng Gu et al.
Journal of chromatography. A, 1376, 53-63 (2014-12-30)
The main objective of our research was to develop silica-based, polymer-functionalized ion exchange materials for single-use bioprocess applications, with the ultimate goal of achieving maximal binding capacity for target proteins. Herein we report the utilization of Grace(®) wide pore silica

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service