Skip to Content
Merck
All Photos(1)

Key Documents

409014

Sigma-Aldrich

Poly(diallyldimethylammonium chloride) solution

20 wt. % in H2O

Synonym(s):

PDADMAC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H16ClN)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

viscous liquid

Quality Level

concentration

20 wt. % in H2O

refractive index

n20/D 1.375

viscosity

60-180 cP(25 °C)

density

1.04 g/mL at 25 °C

SMILES string

[Cl-].[N+](CC=C)(CC=C)(C)C

InChI

1S/C8H16N.ClH/c1-5-7-9(3,4)8-6-2;/h5-6H,1-2,7-8H2,3-4H3;1H/q+1;/p-1

InChI key

GQOKIYDTHHZSCJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Poly(diallyldimethylammonium chloride) solution (PDADMAC) is a cationic polyelectrolyte that is widely utilized as a flocculant. It is prepared by the radical polymerization of diallyldimethylammonium chloride (DADMAC) in presence of a catalyst in the form of an organic peroxide.

Application

PDADMAC can be used as an electrolytic solution in combination with mesoporous silica to form a responsive carrier system for potential usage in drug delivery. It can be also be coated on gold nanorods to enhance the cellular uptake in biomedical systems. It is used in the surface modification of indium tin oxide (ITO) substrate which may be used in chemical sensors or biosensor based applications.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and use of PolyDADMAC for water purification
Biennial Conference of the Water Institute of Southern Africa, Durban, South Africa, 46(8), 2584-2594 (2002)
Synthesis and solution properties of poly (N-isopropylacrylamide-co-diallyldimethylammonium chloride)
Deng Y and Pelton R
Macromolecules, 28(13), 4617-4621 (1995)
Polyelectrolyte multilayer as matrix for electrochemical deposition of gold clusters: toward super-hydrophobic surface
Zhang X, et al.
Journal of the American Chemical Society, 126(10), 3064-3065 (2004)
Lulu Han et al.
Biomaterials, 34(4), 975-984 (2012-11-07)
The directional cell migration plays a crucial role in a variety of physiological and pathological processes. It can be controlled by the gradient cues immobilized on the substrate. The poly(sodium 4-styrenesulfonate) (PSS)/poly(diallyldimethylammonium) chloride (PDADMAC) multilayers were post-treated in a gradient
Solution properties of poly (diallyldimethylammonium chloride)(PDDA)
Marcelo G, et al.
Polymer, 46(8), 2584-2594 (2005)

Articles

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service