Skip to Content
Merck
All Photos(1)

Key Documents

408107

Sigma-Aldrich

[2-(Methacryloyloxy)ethyl]trimethylammonium chloride solution

75 wt. % in H2O

Synonym(s):

2-Trimethylammonioethyl methacrylate chloride, 2-Trimethylammoniumethyl methacrylate chloride, Ethanaminium, N ,N ,N -trimethyl-2-[(2-methyl-1-oxo-2-propenyl)oxy]-chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH2N(CH3)3Cl
CAS Number:
Molecular Weight:
207.70
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

contains

600 ppm monomethyl ether hydroquinone as inhibitor

concentration

75 wt. % in H2O

refractive index

n20/D 1.469

density

1.105 g/mL at 25 °C

SMILES string

[Cl-].CC(=C)C(=O)OCC[N+](C)(C)C

InChI

1S/C9H18NO2.ClH/c1-8(2)9(11)12-7-6-10(3,4)5;/h1,6-7H2,2-5H3;1H/q+1;/p-1

InChI key

RRHXZLALVWBDKH-UHFFFAOYSA-M

Application


  • Multiplexed Electrokinetic Sample Fractionation, Preconcentration and Elution for Proteomics.: The research explores the application of [2-(Methacryloyloxy)ethyl]trimethylammonium chloride in a microfluidic device for efficient sample fractionation and preconcentration in proteomic analyses, demonstrating its utility in enhancing analytical performance (Hua et al., 2013).

  • Fabrication of Positively Charged Poly(ethylene glycol)-Diacrylate Hydrogel as a Bone Tissue Engineering Scaffold.: This paper discusses the integration of [2-(Methacryloyloxy)ethyl]trimethylammonium chloride into a PEG-diacrylate hydrogel, enhancing its suitability as a scaffold in bone tissue engineering due to improved cell adhesion and proliferation properties (Tan et al., 2012).

  • Controlling the Surface Chemistry and Chromatographic Properties of Methacrylate-Ester-Based Monolithic Capillary Columns via Photografting.: This research highlights the role of [2-(Methacryloyloxy)ethyl]trimethylammonium chloride in modifying the surface chemistry of monolithic capillary columns, thereby improving their chromatographic properties for advanced separation techniques (Eeltink et al., 2007).

  • Interaction Between Anionic Dyes and Cationic Flocculant P(AM-DMC) in Synthetic Solutions.: The study investigates the interaction mechanisms between anionic dyes and cationic flocculants, including [2-(Methacryloyloxy)ethyl]trimethylammonium chloride, contributing to the development of more efficient wastewater treatment processes (Shen et al., 2006).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye Tu et al.
International journal of nanomedicine, 11, 5549-5561 (2016-11-09)
We recently reported that electret, which was prepared by a corona charging system with polypropylene film, could enhance the transdermal delivery of several drugs of low molecular weight. The aim of this study was to investigate whether electret could enhance
Katharina Pruessmann et al.
AAPS PharmSciTech, 20(1), 27-27 (2019-01-04)
The aim of this study was to investigate if the geometry of the dissolution vessel, the dissolution medium volume and composition might contribute to the variation in drug release from drug-eluting stents (DES) in different test setups, which has been
Katarzyna Niesyto et al.
Polymers, 12(9) (2020-09-26)
Amphiphilic graft copolymers based on [2-(methacryloyloxy)ethyl]trimethyl- ammonium chloride (TMAMA) were obtained for the delivery of pharmaceutical ionic drugs, such as p-aminosalicylate (PAS) and clavunate (CLV) anions. The side chains were attached by grafting from a multifunctional macroinitiator via atom transfer
Annalisa La Gatta et al.
Journal of functional biomaterials, 3(2), 225-238 (2012-01-01)
Extracellular matrix components such as hyaluronan (HA) and chondroitin sulfate (CS) were combined with a synthetic matrix of p(HEMA-co-METAC) (poly(2-hydroxyethylmethacrylate-co-2-methacryloxyethyltrimethylammonium)) at 1% and 2% w/w concentration following a previously developed procedure. The resulting semi-interpenetrating hydrogels were able to extensively swell
Ying Zhang et al.
Journal of hazardous materials, 340, 309-318 (2017-07-19)
Ongoing research activities are targeted to explore high photocatalytic activity of TiO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service