Skip to Content
Merck
All Photos(2)

Key Documents

325376

Sigma-Aldrich

Bis(2,4-dinitrophenyl) oxalate

98%

Synonym(s):

2,4-Dinitrophenyl oxalate, DNPO, Oxalic acid bis(2,4-dinitrophenyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CO2C6H3(NO2)2]2
CAS Number:
Molecular Weight:
422.22
Beilstein:
2318321
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

mp

189-190 °C (lit.)

functional group

ester
nitro

SMILES string

[O-][N+](=O)c1ccc(OC(=O)C(=O)Oc2ccc(cc2[N+]([O-])=O)[N+]([O-])=O)c(c1)[N+]([O-])=O

InChI

1S/C14H6N4O12/c19-13(29-11-3-1-7(15(21)22)5-9(11)17(25)26)14(20)30-12-4-2-8(16(23)24)6-10(12)18(27)28/h1-6H

InChI key

CBZOGAWUNMFXFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2,4-dinitrophenyl) oxalate (DNPO) is a chemiluminescence reagent.[1] Chemiluminescence produced in the oxidation of DNPO by hydrogen peroxide in the presence of a polycyclic aromatic hydrocarbon was monitored by spectroscopic methods.[2] The kinetics of the imidazole-catalyzed decomposition of DNPO was investigated by the stopped-flow technique.[3]

Application

Bis(2,4-dinitrophenyl) oxalate was used in detection of fluorescent compound dansylalanine in buffer solution by flow injection analysis.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bis (2, 4-dinitrophenyl) oxalate as a chemiluminescence reagent. Determination of fluorescent compounds by flow injection analysis.
Honda K, et al.
Analytical Chemistry, 55(6), 940-943 (1983)
T Kawasaki et al.
Biomedical chromatography : BMC, 4(3), 113-118 (1990-05-01)
Dopamine (DA), norepinephrine (NE) and 3,4-dihydroxybenzylamine (DHBA) are converted to highly fluorescent cyanobenz[f]isoindole (CBI) derivatives by reacting these amines with naphthalene-2,3-dicarboxaldehyde in the presence of cyanide ion. Femtomole amounts of these CBI derivatives separated by reverse-phase high performance liquid chromatography
Andrew G. Hadd et al.
The Journal of organic chemistry, 61(8), 2657-2663 (1996-04-19)
The kinetics of the imidazole-catalyzed decomposition of bis(2,4,6-trichlorophenyl) oxalate (TCPO) and bis(2,4-dinitrophenyl) oxalate (DNPO) was investigated by the stopped-flow technique. Pseudo-first-order rate constants were determined as a function imidazole concentration in the temperature range 6-45 degrees C by fitting the
José Antonio Murillo Pulgarín et al.
Analytica chimica acta, 691(1-2), 76-82 (2011-04-05)
Kinetic and spectroscopic methods were used in combination in this work to develop a new analytical tool for use in chemiluminescence detection processes. Specifically, time-resolved chemiluminescence was used jointly with a stopped-flow assembly in order to monitor the chemiluminescence produced
M B Talawar et al.
Journal of hazardous materials, 124(1-3), 153-164 (2005-06-28)
This paper reviews the recent work done on the synthesis as well as characterization of dinitrogen pentoxide (DNPO). The physico-chemical characteristics of DNPO are also discussed. The review brings out the key aspects of N2O5 technology with relevance to realize

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service