Skip to Content
Merck
All Photos(1)

Key Documents

554073

Sigma-Aldrich

Rubrene

≥98%

Synonym(s):

5,6,11,12-Tetraphenylnaphthacene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H28
CAS Number:
Molecular Weight:
532.67
Beilstein:
1917339
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98%

mp

330-335 °C (lit.)

λmax

299 nm
460 nm (2nd)

SMILES string

c1ccc(cc1)-c2c3ccccc3c(-c4ccccc4)c5c(-c6ccccc6)c7ccccc7c(-c8ccccc8)c25

InChI

1S/C42H28/c1-5-17-29(18-6-1)37-33-25-13-14-26-34(33)39(31-21-9-3-10-22-31)42-40(32-23-11-4-12-24-32)36-28-16-15-27-35(36)38(41(37)42)30-19-7-2-8-20-30/h1-28H

InChI key

YYMBJDOZVAITBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rubrene is a tetraphenyl derivative of tetracene that is used as an organic semiconductor. It is used as a source material in the fabrication of rubrene single crystal based transistors with carrier mobility over 10 cm2V−1s−1.

Application

Reagent for chemiluminescence research and for transition metal complex ligation.
Rubrene based thin films are used as semiconducting layers along with poly(vinylidene fluoride-terefluoroethylene) (PVDF-TeFE) as insulating layers for the fabrication of ferro-electric gate field effect transistors (FETs). It may also be used as high mobility transporting material in the development of single crystal FETs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High performance of rubrene thin film transistor by weak epitaxy growth method
Chang H, et al.
Organic Electronics, 20, 43-48 (2015)
Organic ferroelectric gate field-effect transistor memory using high-mobility rubrene thin film
Kanashima T, et al.
Japanese Journal of Applied Physics, 53(4S), 04ED11-04ED11 (2014)
Intrinsic charge transport on the surface of organic semiconductors
Podzorov V, et al.
Physical Review Letters, 93(8), 086602-086602 (2004)
Field-effect transistors on rubrene single crystals with parylene gate insulator
Podzorov V, et al.
Applied Physics Letters, 82(11), 1739-1741 (2003)
Influence of the gate dielectric on the mobility of rubrene single-crystal field-effect transistors
Stassen AF, et al.
Applied Physics Letters, 85(17), 3899-3901 (2004)

Articles

In this article, we demonstrate that bis-styrylbenzene derivatives show promising characteristics for very low lasing thresholds and discuss the design considerations for organic lasing molecules.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service