All Photos(1)
About This Item
Linear Formula:
Ce(OH)4
CAS Number:
Molecular Weight:
208.15
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
chunks
Recommended Products
grade
for analytical purposes
form
chunks
reaction suitability
reagent type: catalyst
core: cerium
concentration
≥47.1% Ce (after ignition to oxide, gravimetric)
SMILES string
O[Ce](O)(O)O
InChI
1S/Ce.4H2O/h;4*1H2/q+4;;;;/p-4
InChI key
WTVAYLQYAWAHAX-UHFFFAOYSA-J
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Fumihiko Ogata et al.
Journal of oleo science, 60(3), 133-138 (2011-02-24)
In the present study, the phosphate adsorption abilities of granular materials, gibbsite (GB) and cerium hydroxide (CE), granulated with ethyl cellulose and ethanol, were investigated. For suitable granular conditions using GB or CE, the percentage of binder and amount of
A Vazquez-Torres et al.
Science (New York, N.Y.), 287(5458), 1655-1658 (2000-03-04)
A type III protein secretion system encoded by Salmonella pathogenicity island 2 (SPI2) has been found to be required for virulence and survival within macrophages. Here, SPI2 was shown to allow Salmonella typhimurium to avoid NADPH oxidase-dependent killing by macrophages.
G S Groenewold et al.
Physical chemistry chemical physics : PCCP, 9(5), 596-606 (2007-01-24)
Cerium(iii) hydroxy reactive sites are responsible for several important heterogeneous catalysis processes, and understanding the reaction chemistry of substrate molecules like CO, H(2)O, and CH(3)OH as they occur in heterogeneous media is a challenging task. We report here the first
G Nakos et al.
Acta histochemica, 95(2), 203-219 (1993-12-01)
H2O2-generating monoamine oxidase can be visualized in the light microscope with tetrazolium, metal salt (ferricyanide) and coupled peroxidatic oxidation methods. Due to methodological draw-backs these procedures do no allow satisfactory results. In search for an alternative method a light microscopic
Umit Divrikli et al.
Environmental monitoring and assessment, 138(1-3), 167-172 (2007-06-15)
A method for the preconcentration of the total chromium based on coprecipitation with cerium (IV) hydroxide is proposed for determination of chromium by flame atomic absorption spectrometry. Different factors including carrier element amount, pH, sample volume and matrix ion effects
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service