Skip to Content
Merck
All Photos(3)

Key Documents

289787

Sigma-Aldrich

Copper(II) hydroxide

technical grade

Synonym(s):

Cupric hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(OH)2
CAS Number:
Molecular Weight:
97.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
technical grade
form:
powder
solubility:
H2O: insoluble(lit.)
aqueous acid: slightly soluble(lit.)
Pricing and availability is not currently available.

grade

technical grade

form

powder

contains

stabilizer

concentration

≥57.0% Cu (EDTA titration)

solubility

H2O: insoluble(lit.)
aqueous acid: slightly soluble(lit.)

application(s)

battery manufacturing

SMILES string

O[Cu]O

InChI

1S/Cu.2H2O/h;2*1H2/q+2;;/p-2

InChI key

JJLJMEJHUUYSSY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

The orthorhombic nature of copper hydroxide crystals was determined by X ray diffraction. [1] Copper hydroxide can act as a heterogeneous catalyst in the selective oxidative cross coupling of terminal alkynes to yield their corresponding ynamides. [2]

Application

Copper hydroxide based monoliths can be used in the synthesis of copper hydroxide-based monolithic xerogels. Potential applications of this metal organic frameworks (MOFs) include gas storage, separation, drug delivery, and biomedicine. [3][4] Supported Cu(OH)x can be used as a catalyst for the aerobic cross dehydrogenative coupling of benzenethiols and cyclic amides to yield N-acylsulfenamides. [5]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sonja Kosuta et al.
Journal of environmental quality, 31(3), 910-916 (2002-05-25)
The City of Montreal, Canada, evaluated the environmental impact and usefulness of in-ground copper (Cu)-treated baskets in controlling root growth of hardwood trees in nursery culture. Using baskets planted with 5-yr-old Norway maple (Acer platanoides L.) trees, the amount and
E S Vasetskiĭ et al.
Izvestiia Akademii nauk SSSR. Seriia biologicheskaia, (1)(1), 148-151 (1989-01-01)
A specific cosedimentation of proteins and their complexes with DNA at low temperature (M-band technique) has been demonstrated. Model experiments with reconstituted SV40 DNA-topoisomerase I and SV40 DNA-E. coli RNA polymerase complexes demonstrated the potential and capacities of the method.
Mechanically stable, hierarchically porous Cu 3 (btc) 2 (HKUST-1) monoliths via direct conversion of copper (ii) hydroxide-based monoliths.
Moitra N, et al.
Chemical Communications (Cambridge, England), 51(17), 3511-3514 (2015)
Heterogeneously catalyzed selective aerobic oxidative cross-coupling of terminal alkynes and amides with simple copper (ii) hydroxide.
Jin X, et al.
Chemical Communications (Cambridge, England), 48(41), 4974-4976 (2012)
Caroline K Nguyen et al.
Water research, 45(16), 5302-5312 (2011-08-27)
A rapid reaction between free chlorine and the cupric hydroxide [Cu(OH)2] solids commonly found on pipe walls in premise plumbing can convert free chlorine to chloride and rapidly age Cu(OH)2 to tenorite (CuO). This reaction has important practical implications for

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service