Skip to Content
Merck
All Photos(4)

Key Documents

291234

Sigma-Aldrich

Dibutyltin dilaurate

95%

Synonym(s):

DBTDL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2Sn[OCO(CH2)10CH3]2
CAS Number:
Molecular Weight:
631.56
Beilstein:
4156980
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.2 mmHg ( 160 °C)

Quality Level

Assay

95%

refractive index

n20/D 1.471 (lit.)

density

1.066 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC(=O)O[Sn](CCCC)(CCCC)OC(=O)CCCCCCCCCCC

InChI

1S/2C12H24O2.2C4H9.Sn/c2*1-2-3-4-5-6-7-8-9-10-11-12(13)14;2*1-3-4-2;/h2*2-11H2,1H3,(H,13,14);2*1,3-4H2,2H3;/q;;;;+2/p-2

InChI key

UKLDJPRMSDWDSL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyltin dilaurate, also known as dibutyltin didodecanoate, is a dibutyltin laurate hydroxide intermediate that is commonly used as a catalyst. It is also used in the transesterification reaction. [1]

Application

Dibutyltin dilaurate is used as a catalyst in:
  • The synthesis of polyurethane (PU) and polyhydroxyurethane (PHU) prepolymers and lignin urethane. [2] [3]
  • A protocol for the covalent attachment of poly(ethylene glycol) (PEG) to silicon oxide to form a hydrophilic non-fouling surface.[4]
  • Preparation of polymers by reacting hydroxyl-terminated macromonomers and aliphatic diisocyanates.[5][6][7][8]

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Sens. 1 - STOT RE 1 - STOT SE 1

Target Organs

thymus, thymus,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

372.2 - 379.4 °F - closed cup

Flash Point(C)

189 - 193 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derivatization of pillared silicon substrates using poly (ethylene glycol) and 1-dodecanethiol.
White A
Journal of undergraduate neuroscience education : JUNE : a publication of FUN, Faculty for Undergraduate Neuroscience, 7(1), 91-98 (2007)
Mei Li et al.
ACS omega, 5(1), 529-536 (2020-01-21)
A tung-oil-based polyphenol (ATOM), containing the phenolic hydroxyl group, was synthesized from tung oil and 4-maleimidophenol by the Diels-Alder addition reaction. Then self-healing thermosetting polyurethanes were prepared from ATOM and the polyurethane prepolymer. The chemical structure and cross-link network were
High bio-content polyurethane composites with urethane modified lignin as filler
Chaoqun Z,et al.
Polymer, 69, 52-57 (2015)
Preparation of polyurethane/acrylic hybrid nanoparticles via a miniemulsion polymerization process.
Li M
Macromolecules, 38(10), 4183-4192 (2005)
Self-assembled supramolecular hydrogels formed by biodegradable PEO?PHB?PEO triblock copolymers and ?-cyclodextrin for controlled drug delivery.
Li J
Biomaterials, 27(22), 4132-4140 (2006)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the concentration of item 291234?

    1 answer
    1. The concentration (Molarity) of item 291234 can be calculated using the formula: Density x Volume x Concentration / Molecular weight = Molarity. For example, if the density is 1.066g, the volume is 1000, and the molecular weight is 631.59, the resulting molarity would be 1.69M.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service