Skip to Content
Merck
All Photos(3)

Key Documents

256439

Sigma-Aldrich

4,4′-Methylenebis(phenyl isocyanate)

98%

Synonym(s):

4,4′-MDI, Bis(4-isocyanatophenyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(C6H4NCO)2
CAS Number:
Molecular Weight:
250.25
Beilstein:
797662
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

reaction suitability

reagent type: cross-linking reagent

bp

200 °C/5 mmHg (lit.)

mp

42-45 °C (lit.)

density

1.18 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

O=C=Nc1ccc(Cc2ccc(cc2)N=C=O)cc1

InChI

1S/C15H10N2O2/c18-10-16-14-5-1-12(2-6-14)9-13-3-7-15(8-4-13)17-11-19/h1-8H,9H2

InChI key

UPMLOUAZCHDJJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Methylenebis(phenyl isocyanate) (MDI) is an aromatic diisocyanates class of monomer that is widely used in the production of polyurethane plastics, foam insulation, coatings, adhesives, and sealants. It is highly reactive due to the presence of two isocyanate functional groups. MDI is known for its excellent strength, durability, and resistance to chemical and environmental damage, which makes it useful in resin composition, lithographic printing plates, coating films, optical films, image display devices, semiconductor devices, and polyurethane foam production.

Application

4,4′-Methylenebis(phenyl isocyanate) can be used as a starting material to synthesize:
  • Polyurethane cationomers, that are applicable in medical implants.
  • A prepolymer for preparing self-healable polyurethane elastomers.
It can also be used as a cross-linking agent to synthesize PEBA (polyether-block-amide) copolymer with improved mechanical properties, which are mainly used in soles, medical tubes, aerospace parts, and chemical separation. Additionally, MDI is also used as a crosslinker to covalently modify graphene oxide (GO) and enhance the corrosion resistance of polystyrene coatings. The resultant polystyrene/GO-MDI composite coatings showed superior corrosion resistance to unmodified polystyrene coatings. This approach can potentially be used to improve the durability of materials in various industrial and biomedical applications.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

411.8 °F - closed cup

Flash Point(C)

211 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jyun-Yan Ye et al.
Materials (Basel, Switzerland), 14(4) (2021-02-11)
A series of N-substituted polyether-block-amide (PEBA-X%) copolymers were prepared by melt polycondensation of nylon-6 prepolymer and polytetramethylene ether glycol at an elevated temperature using titanium isopropoxide as a catalyst. The structure, thermal properties, and crystallinity of PEBA-X% were investigated using
Journal of Applied Physiology, 70, 6983-6983 (1991)
Preparation and Characterization of Isosorbide-Based Self-Healable Polyurethane Elastomers with Thermally Reversible Bonds
Han-Na Kim, et al.
Molecules (Basel), 24 (2019)
Dominika Swierczyńska-Machura et al.
Medycyna pracy, 62(6), 567-577 (2012-02-09)
The aim of the study was to analyze the data obtained during the diagnostic process and their real values in the final diagnosis of patients exposed to diisocyanates amd suspected of occupational allergic disease of the respiratory system The patients
Unique foreign body ingestions in the pediatric population.
Sheila Isabelle Goertemoeller et al.
Journal of emergency nursing: JEN : official publication of the Emergency Department Nurses Association, 36(2), 178-179 (2010-03-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service