Skip to Content
Merck
All Photos(3)

Key Documents

A58003

Sigma-Aldrich

2-Amino-4-hydroxy-6-methylpyrimidine

98%

Synonym(s):

2-Amino-6-methyl-4-pyrimidinol, 6-Methylisocytosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3O
CAS Number:
Molecular Weight:
125.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

>300 °C (lit.)

SMILES string

Cc1cc(O)nc(N)n1

InChI

1S/C5H7N3O/c1-3-2-4(9)8-5(6)7-3/h2H,1H3,(H3,6,7,8,9)

InChI key

KWXIPEYKZKIAKR-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuande Xu et al.
Journal of biomaterials science. Polymer edition, 31(6), 712-728 (2020-01-21)
Gelatin-UPy based on gelatin with ureidopyrimidinone (UPy) side chains was prepared with varying content of UPy units. On increasing the UPy content, the glass transition temperature, crystallinity and swelling decreased. Gelatin-UPy demonstrated self-healing properties as the UPy units could reversibly
Joong Tark Han et al.
Scientific reports, 7(1), 4931-4931 (2017-07-12)
Most synthetic processes of metallic nanostructures were assisted by organic/inorganic or polymeric materials to control their shapes to one-dimension or two-dimension. However, these additives have to be removed after synthesis of metal nanostructures for applications. Here we report a straightforward
Jérôme Mulhbacher et al.
PLoS pathogens, 6(4), e1000865-e1000865 (2010-04-28)
Riboswitches are regulatory elements modulating gene expression in response to specific metabolite binding. It has been recently reported that riboswitch agonists may exhibit antimicrobial properties by binding to the riboswitch domain. Guanine riboswitches are involved in the regulation of transport
Bo Li et al.
Nanoscale, 11(2), 467-473 (2018-12-20)
Supramolecular polymers show unique and excellent properties due to the reversible and designable nature of the non-covalent interactions. Herein, ureido-pyrimidinone (UPy)-based supramolecular polymers were employed to fabricate the thermo-responsive composite materials with multi-walled carbon nanotubes (MWCNTs) by planting the MWCNTs
Jingsi Chen et al.
ACS applied materials & interfaces, 11(20), 18720-18729 (2019-05-03)
Although recent years have witnessed intense efforts and innovations in the design of flexible conductive materials for the development of next-generation electronic devices, it remains a great challenge to integrate multifunctionalities such as stretchability, self-healing, adhesiveness, and sensing capability into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service