Skip to Content
Merck
All Photos(1)

Key Documents

284246

Sigma-Aldrich

Nonacosane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)27CH3
CAS Number:
Molecular Weight:
408.79
Beilstein:
1724922
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

286 °C/15 mmHg (lit.)

mp

63-66 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C29H60/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-29H2,1-2H3

InChI key

IGGUPRCHHJZPBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonacosane is a major paraffin of most plant waxes.The metabolic fate of n-nonacosane (from cabbage wax) in Micrococcus cerificans, a soil organism has been investigated. Solubility of solid n-nonacosane in supercritical ethane has been reported.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubilities of solid n-nonacosane and n-tritriacontane in supercritical ethane.
Moradinia I and Teja AS.
Journal of Chemical and Engineering Data, 33(3), 240-242 (1988)
Metabolism of a plant wax paraffin (n-nonacosane) in the rat.
P E Kolattukudy et al.
The Journal of nutrition, 90(2), 167-174 (1966-10-01)
Metabolism of a plant wax paraffin (n-nonacosane) by a soil bacterium (Micrococcus cerificans).
L Hankin et al.
Journal of general microbiology, 51(3), 457-463 (1968-05-01)
[Constituents of the Lipophilic fraction of the Apocynum venetum leaf].
P F Jiang
Zhong yao tong bao (Beijing, China : 1981), 10(5), 30-31 (1985-05-01)
Eoin P Coll et al.
The journal of physical chemistry. B, 111(30), 8702-8704 (2007-07-05)
Plant wax contains long-chain alkanes and related components which are transported to the surface of the plant by specialized ABC transporter proteins. Here, we determine the distribution and conformation of three wax components, nonacosane, nonacosan-15-one, and nonacosan-15-ol, using unbiased and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service