Skip to Content
Merck
All Photos(1)

Key Documents

263850

Sigma-Aldrich

Tricosane

99%

Synonym(s):

n-Tricosane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)21CH3
CAS Number:
Molecular Weight:
324.63
Beilstein:
1756000
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

199-200 °C/3 mmHg (lit.)

mp

46-47 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C23H48/c1-3-5-7-9-11-13-15-17-19-21-23-22-20-18-16-14-12-10-8-6-4-2/h3-23H2,1-2H3

InChI key

FIGVVZUWCLSUEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tricosane is an aliphatic alkyl chain that can be used:
  • As a phase change material for electronic cooling applications.
  • To prepare ticosane functionalized single-walled carbon nanotube (SWCNT) biosensors for the detection of volatile organic molecules in human breath.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Heat pipe with PCM for electronic cooling
Weng Y-C, et al.
Applied Energy, 88(5), 1825-1833 (2011)
Disorder in the pseudohexagonal rotator phase of n-alkanes: molecular-dynamics calculations for tricosane.
Ryckaert J-P, et al.
Molecular Physics, 67(5), 957-979 (1989)
Experimental study of thermal energy storage characteristics using heat pipe with nano-enhanced phase change materials
Krishna J, et al.
IOP Conference Series: Materials Science and Engineering, 225, 012058-012058 (2017)
Enrique Blázquez-Blázquez et al.
Polymers, 12(6) (2020-06-18)
A detailed study of the phase behavior of n-paraffin C23H48 has been performed by means of real-time variable-temperature experiments with synchrotron radiation. Two detectors were employed for simultaneous analysis of the small-angle (SAXS) and wide-angle X-ray-scattering (WAXS) regions. This paraffin
Single-walled carbon nanotube-based biosensors for the detection of volatile organic compounds of lung cancer
Liu FL, et al.
Physica E: Low-Dimensional Systems and Nanostructures, 44(2), 367-372 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service