Skip to Content
Merck
All Photos(3)

Key Documents

134457

Sigma-Aldrich

Docosane

99%

Synonym(s):

n-Docosane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)20CH3
CAS Number:
Molecular Weight:
310.60
Beilstein:
1702206
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

10.8 (vs air)

vapor pressure

<1 mmHg ( 21.1 °C)

Assay

99%

form

solid

bp

369 °C (lit.)

mp

42-45 °C (lit.)

density

0.778 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C22H46/c1-3-5-7-9-11-13-15-17-19-21-22-20-18-16-14-12-10-8-6-4-2/h3-22H2,1-2H3

InChI key

HOWGUJZVBDQJKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Docosane was used to investigate commercially available waxes in the form of thin disc samples as possible diffraction intensity standards for macromolecular crystallography synchrotron beamlines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Brandao-Neto et al.
Journal of synchrotron radiation, 17(1), 53-60 (2009-12-24)
A number of commercially available waxes in the form of thin disc samples have been investigated as possible diffraction intensity standards for macromolecular crystallography synchrotron beamlines. Synchrotron X-ray powder diffraction measurements show that beeswax offers the best performance of these
Leisha M Armijo et al.
Journal of nanobiotechnology, 18(1), 35-35 (2020-02-20)
Novel methods are necessary to reduce morbidity and mortality of patients suffering from infections with Pseudomonas aeruginosa. Being the most common infectious species of the Pseudomonas genus, P. aeruginosa is the primary Gram-negative etiology responsible for nosocomial infections. Due to
Auke J Kronemeijer et al.
Small (Weinheim an der Bergstrasse, Germany), 7(11), 1593-1598 (2011-05-04)
Charge transport through alkanes and para-phenylene oligomers is investigated in large-area molecular junctions. The molecules are self-assembled in a monolayer and contacted with a top electrode consisting of poly(3,4-ethylenedioxythiophene)-poly(4-styrenesulfonic acid) (PEDOT:PSS). The complete set of J(V,T) characteristics of both saturated
Cédric Pisani et al.
Nanoscale, 9(5), 1840-1851 (2016-11-20)
Magnetic mesoporous silica nanoparticles (M-MSNs) represent promising targeting tools for theranostics. Engineering the interaction of nanoparticles (NPs) with biological systems requires an understanding of protein corona formation around the nanoparticles as this drives the biological fate of nanocarriers. We investigated
H Kraack et al.
The Journal of chemical physics, 121(16), 8003-8009 (2004-10-16)
The temperature dependent phase behavior of Langmuir films of n-alkanes [CH3(CH2)(n-2)CH3, denote Cn] on mercury was studied for chain lengths 19< or =n< or =22 and temperatures 15< or =T< or =44 degrees C, using surface tensiometry and surface x-ray

Protocols

Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service