Skip to Content
Merck
All Photos(3)

Key Documents

370533

Sigma-Aldrich

1H,1H,2H,2H-Perfluoro-1-octanol

97%

Synonym(s):

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 6:2 FTOH, 1H,1H,2H,2H-Perfluoro-1-octanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)5CH2CH2OH
CAS Number:
Molecular Weight:
364.10
Beilstein:
1992757
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

refractive index

n20/D 1.313 (lit.)

bp

88-95 °C/28 mmHg (lit.)

density

1.651 g/mL at 25 °C (lit.)

SMILES string

OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C8H5F13O/c9-3(10,1-2-22)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h22H,1-2H2

InChI key

GRJRKPMIRMSBNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1H,1H,2H,2H-Perfluoro-1-octanol is a fluorotelomer alcohol.[1]

Application

1H,1H,2H,2H-Perfluoro-1-octanol may be used to generate droplets in droplet-based microfluidics systems.[2][3] It can be used as a starting material in the synthesis of 1H,1H,2H,2H -perfluorooctyl vinyl ether (FOVE).[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Berna Özkale et al.
Lab on a chip, 19(5), 778-788 (2019-02-05)
We present a methodology for building biologically inspired, soft microelectromechanical systems (MEMS) devices. Our strategy combines several advanced techniques including programmable colloidal self-assembly, light-harvesting with plasmonic nanotransducers, and in situ polymerization of compliant hydrogel mechanisms. We synthesize optomechanical microactuators using
Max M Villa et al.
mSystems, 5(3) (2020-07-02)
Culture and screening of gut bacteria enable testing of microbial function and therapeutic potential. However, the diversity of human gut microbial communities (microbiota) impedes comprehensive experimental studies of individual bacterial taxa. Here, we combine advances in droplet microfluidics and high-throughput
Non-spherical gelatin particle in two phases microfluidic system.
Simone G
Microelectronic Engineering, 111, 339-342 (2013)
Living cationic polymerization of 1H, 1H, 2H, 2H perfluorooctyl vinyl ether.
Vandooren C
Polymer Bull., 32(4), 387-393 (1994)
Dong Chen et al.
ACS nano, 11(12), 11978-11985 (2017-12-05)
Emulsions of two immiscible liquids can slowly coalesce over time when stabilized by surfactant molecules. Pickering emulsions stabilized by colloidal particles can be much more stable. Here, we fabricate biocompatible amphiphilic dimer particles using a hydrogel, a strongly hydrophilic material

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service