Skip to Content
Merck
All Photos(2)

Key Documents

271993

Sigma-Aldrich

2,5-Dibromo-6-isopropyl-3-methyl-1,4-benzoquinone

Synonym(s):

2,5-Dibromo-3-methyl-6-isopropyl-p-benzoquinone, Dibromothymoquinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6Br2(CH3)(=O)2
CAS Number:
Molecular Weight:
321.99
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

form

powder, crystals or chunks

Quality Level

mp

70-72 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(C)C1=C(Br)C(=O)C(C)=C(Br)C1=O

InChI

1S/C10H10Br2O2/c1-4(2)6-8(12)9(13)5(3)7(11)10(6)14/h4H,1-3H3

InChI key

GHHZELQYJPWSMG-UHFFFAOYSA-N

Application

2, 5-Dibromo-6-isopropyl-3-methyl-1, 4-benzoquinone has been used as a metabolic inhibitor of violaxanthin (V) cycle of Pelvetia canaliculata and is used as a photosynthetic electron transport chain (pETC) inhibitor.[1][2]

Biochem/physiol Actions

2, 5-Dibromo-6-isopropyl-3-methyl-1, 4-benzoquinone acts as an inhibitor of respiratory and photosynthetic processes.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sousuke Imamura et al.
FEBS letters, 554(3), 357-362 (2003-11-19)
The expression of group 2 sigma factors is characterized in a cyanobacterium Synechocystis sp. PCC 6803 grown in culture, changing light conditions (white, red and blue light, and darkness), or the presence of drugs (rifampicin, chloramphenicol, DCMU, and DBMIB), and
Deepak Venkanna et al.
Frontiers in plant science, 8, 1347-1347 (2017-08-22)
The protein superfamily of short-chain dehydrogenases/reductases (SDR), including members of the atypical type (aSDR), covers a huge range of catalyzed reactions and
Yuuki Sakai et al.
Planta, 221(6), 823-830 (2005-04-06)
In epidermal cells of the aquatic angiosperm Vallisneria gigantea Graebner, high-intensity blue light (BL) induces the avoidance response of chloroplasts. We examined simultaneous BL-induced changes in the configuration of actin filaments in the cytoplasmic layers that face the outer periclinal
Shan Gao et al.
Journal of experimental botany, 63(12), 4349-4358 (2012-03-23)
Porphyra yezoensis, a representative species of intertidal macro-algae, is able to withstand periodic desiccation at low tide but is submerged in seawater at high tide. In this study, changes in photosynthetic electron flow in P. yezoensis during desiccation and re-hydration
Ireneusz Slesak et al.
Plant & cell physiology, 44(6), 573-581 (2003-06-27)
Mesembryanthemum crystallinum, a facultative halophyte and C(3)-Crassulacean acid metabolism (CAM) intermediate plant, has become a favoured plant for studying stress response mechanisms during C(3)-CAM shifts. One hour of exposure to excess light (EL) caused inhibition of photosynthetic electron transport in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service