Skip to Content
Merck
All Photos(1)

Documents

252387

Sigma-Aldrich

Pentafluoropropionic anhydride

99%

Synonym(s):

PFPA, Perfluoropropionic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CF2CO)2O
CAS Number:
Molecular Weight:
310.05
Beilstein:
1806446
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

bp

69-70 °C/735 mmHg (lit.)

density

1.571 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)F

InChI

1S/C6F10O3/c7-3(8,5(11,12)13)1(17)19-2(18)4(9,10)6(14,15)16

InChI key

XETRHNFRKCNWAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pentafluoropropionic anhydride was used:
  • in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid
  • as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection
  • in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kohno et al.
Biochemical and biophysical research communications, 140(1), 448-454 (1986-10-15)
This is the first report to identify 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain. The detection was performed by coupled gas chromatography - multiple ion detection, using heptafluorobutyric anhydride and pentafluoropropionic anhydride as derivatizing reagents. These amines
Simultaneous determination of 3,4-dihydroxyphenylacetic acid and homovanillic acid in milligram amounts of rat striatal tissue by gas-liquid chromatography.
E Watson et al.
Life sciences, 15(12), 2167-2178 (1974-12-15)
Derivatization and gas chromatographic determination of some biologically important acids in cerebrospinal fluid.
E Watson et al.
Analytical biochemistry, 59(2), 441-451 (1974-06-01)
E Ballesteros et al.
Analytical chemistry, 65(13), 1773-1778 (1993-07-01)
A new analytical method that combines on-line derivatization-extraction and gas-liquid chromatography for the determination of N-methylcarbamates is reported. The hydrolysis products of N-methylcarbamates (phenols) are derivatized and extracted in a continuous fashion using pentafluoropropionic anhydride (derivatizing reagent) in n-hexane as
C Jurado et al.
Forensic science international, 145(2-3), 161-166 (2004-09-29)
This paper describes a procedure for the detection and quantification of ethyl-glucuronide (EtG) in hair samples. During method development the efficacy of extraction of EtG from hair was compared in four extraction methods: (a) methanol; (b) methanol:water (1:1); (c) water;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service