Skip to Content
Merck
All Photos(2)

Key Documents

262242

Sigma-Aldrich

1,4-Phenylene diisocyanate

Synonym(s):

1,4-Diisocyanatobenzene, p-Phenylene diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NCO)2
CAS Number:
Molecular Weight:
160.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.01 mmHg ( 20 °C)

Quality Level

form

solid

bp

260 °C (lit.)

mp

96-99 °C (lit.)

solubility

THF: soluble
acetone: soluble
ethyl acetate: soluble
toluene: soluble

functional group

isocyanate

SMILES string

O=C=Nc1ccc(cc1)N=C=O

InChI

1S/C8H4N2O2/c11-5-9-7-1-2-8(4-3-7)10-6-12/h1-4H

InChI key

ALQLPWJFHRMHIU-UHFFFAOYSA-N

Related Categories

Application

1,4-Phenylene diisocyanate (PPDI) can be used as:
  • A reactant to synthesize high yield polymers via criss-cross addition polymerization reaction with alkyl aldazines in pyridine.
  • An organic building block to prepare adamantyl-containing diurea and thiourea derivatives applicable as potent mammalian and human soluble epoxide hydrolase inhibitors.
  • A coupling agent to synthesize polyethylene glycol (PEG)/silica composite material using dibutyltin dilaurate (DBTDL) as a catalyst.

It can also be used:
  • As a cross-linking reagent to investigate the mechanism of enzyme immobilization on silanized surfaces.
  • In the synthesis of the dipodal bis-urea receptor, a selective receptor for hydrogen sulfate.
  • In the deposition of polyurea resists films via molecular layer deposition.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nesrine Aissaoui et al.
Langmuir : the ACS journal of surfaces and colloids, 30(14), 4066-4077 (2014-03-19)
We investigated the mechanism of enzyme immobilization on silanized surfaces through coupling agents (cross-linkers) in order to understand the role of these molecules on interfacial processes and their effect on catalytic activity. To this end, we used a model multimeric
Grafting of polyethylene glycols onto nanometer silica surface by 1, 4-phenylene diisocyanate
Zhang W and Lee Hong-Ro
Surface and Interface Analysis : SIA, 42(9), 1495-1498 (2010)
Crisscross addition polymerization of alkyl aldazines and 1, 4-phenylene diisocyanate
Hashidzume A, et al.
Polymer, 50(10), 2246-2251 (2009)
S K Dogan et al.
Journal of the mechanical behavior of biomedical materials, 71, 349-361 (2017-04-14)
Poly(lactic acid) (PLA)/thermoplastic polyurethane (TPU) blends were melt-mixed and compatibilized to investigate their biocompatibility, biodegradability and thermally induced shape memory properties. The blend compositions were PLA/TPU: 80/20 (20TPU) and PLA/TPU: 50/50 (50TPU). 1,4-phenylene diisocyanate (PDI) was used in order to
Han Zhou et al.
ACS applied materials & interfaces, 3(2), 505-511 (2011-02-10)
Photoresist materials comprise one of the main challenges faced by lithography to meet the requirements of electronic device size scaling. Here we report for the first time the use of molecular layer deposition (MLD) to produce photoresist materials with controllable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service