Skip to Content
Merck
All Photos(1)

Documents

223867

Sigma-Aldrich

Tolylene-2,6-diisocyanate

97%

Synonym(s):

2-Methyl-m-phenylene diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NCO)2
CAS Number:
Molecular Weight:
174.16
Beilstein:
2211546
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

autoignition temp.

>1148 °F

refractive index

n20/D 1.571 (lit.)

bp

129-133 °C/18 mmHg (lit.)

density

1.225 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1c(cccc1N=C=O)N=C=O

InChI

1S/C9H6N2O2/c1-7-8(10-5-12)3-2-4-9(7)11-6-13/h2-4H,1H3

InChI key

RUELTTOHQODFPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Carbonnelle et al.
The Analyst, 121(5), 663-669 (1996-05-01)
In recent years, epidemiological evidence that exposure to toluene diisocyanate (TDI) is associated with adverse health effects has led to the development of useful analytical methods for the biological monitoring of TDI. In this paper, an HPLC method is presented
G Sesana et al.
American Industrial Hygiene Association journal, 52(5), 183-186 (1991-05-01)
A new sampling system is presented that uses solid sorbent media contained in a tube for the determination of airborne isocyanates (2.4-2.6 toluene diisocyanate, hexamethylene diisocyanate, and 4.4' diaminodiphenylmethane diisocyanate). The method is compared with the National Institute for Occupational
J Huang et al.
Archives of toxicology, 67(6), 373-378 (1993-01-01)
Groups of guinea pigs were exposed via inhalation to toluene diisocyanate (TDI) ranging from 0.02 to 1.0 ppm for 3 h/day on 5 consecutive days. Three weeks later, guinea pigs were challenged with TDI-GSA conjugates. Evaluations were based on TDI
Justin M Hettick et al.
Analytical biochemistry, 414(2), 232-238 (2011-04-05)
Diisocyanates are highly reactive chemical compounds widely used in the manufacture of polyurethanes. Although diisocyanates have been identified as causative agents of allergic respiratory diseases, the specific mechanism by which these diseases occur is largely unknown. To better understand the
Hui-Jung Yeh et al.
Journal of applied toxicology : JAT, 28(2), 189-195 (2007-06-22)
Toluene diisocyanates (TDI) are commonly used in polyurethane (PU)-related products. TDIs have been documented as the leading cause of occupational asthma. Skin exposure to TDI in the workplace is common. However, no studies in the literature have investigated the exact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service