Skip to Content
Merck
All Photos(1)

Key Documents

255270

Sigma-Aldrich

o-Cymene

98%

Synonym(s):

1-Isopropyl-2-methylbenzene, 2-Isopropyltoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(CH3)2
CAS Number:
Molecular Weight:
134.22
Beilstein:
1850838
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

3.6 mmHg ( 37.7 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

178 °C (lit.)

mp

−72-−71 °C (lit.)

solubility

organic solvents: miscible(lit.)
water: insoluble(lit.)

density

0.877 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1ccccc1C

InChI

1S/C10H14/c1-8(2)10-7-5-4-6-9(10)3/h4-8H,1-3H3

InChI key

WWRCMNKATXZARA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Intact and direct dealkylating nitration of o-cymene with nitronium tetronium tetrafluoroborate has been reported. The kinetics of isomerization of cymene has been studied.

Application

  • Mechanism of β–cyclodextrin-thyme nanocomplex formation and release: Explores the encapsulation and release mechanisms of o-Cymene in nanocomplexes, suggesting its potential applications in food industry innovations (Dorofte et al., 2024).
  • Processing of sapodilla (Manilkara zapota L.) fruit: Reports the use of o-Cymene in improving the shelf life of sapodilla fruit, illustrating its role in food preservation and post-harvest loss control (Kumar et al., 2024).
  • Comparative Study on Insecticidal Activities of Essential Oils: Highlights the effectiveness of o-Cymene in essential oil formulations used as insecticides, providing insights into its use in sustainable pest management (Awojide et al., 2023).
  • Chemical composition of essential oil from Lindera caesia: Examines the antifungal and antibiofilm properties of o-Cymene in essential oils, suggesting potential applications in healthcare, especially in dental caries prevention (Zaini et al., 2023).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huijuan Zhang et al.
Food chemistry, 289, 340-350 (2019-04-09)
Dried tangerine peel (DTP) is an excellent plant resource that has been used as ingredients for both food and traditional Chinese medicine. In this study, the efficiency of four different dietary preparation methods (i.e. soaking, boiling, steaming, and ethanol extraction)
Peter K Njuguna et al.
Journal of agricultural and food chemistry, 66(32), 8504-8513 (2018-07-26)
The main hosts of the melon fly Zeugodacus cucurbitate are cultivated and wild cucurbitaceous plants. In eastern Africa, the melon fly is a major pest of the Solanaceae plant Solanum lycopersicum (tomato). We hypothesized that shared species-specific volatiles may play
Alexandros Patsilinakos et al.
Molecules (Basel, Switzerland), 24(5) (2019-03-06)
Biofilm resistance to antimicrobials is a complex phenomenon, driven not only by genetic mutation induced resistance, but also by means of increased microbial cell density that supports horizontal gene transfer across cells. The prevention of biofilm formation and the treatment
[Toxicologic properties and the nature of the combined effect of a orthomenthane hydrocarbon mixture].
V A Stel'makh
Gigiena truda i professional'nye zabolevaniia, (9)(9), 34-37 (1988-09-01)
Kinetics of Three-compound Equilibrations. I. The Isomerization of Cymene1.
Allen RH, et al.
Journal of the American Chemical Society, 81(1), 42-46 (1959)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service