Skip to Content
Merck
All Photos(3)

Key Documents

200018

Sigma-Aldrich

2,3-Dihydrofuran

99%

Synonym(s):

2,3-DHF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O
CAS Number:
Molecular Weight:
70.09
Beilstein:
103168
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

14.46 psi ( 55 °C)
3.67 psi ( 20 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.423 (lit.)

bp

54-55 °C (lit.)

density

0.927 g/mL at 25 °C (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

C1CC=CO1

InChI

1S/C4H6O/c1-2-4-5-3-1/h1,3H,2,4H2

InChI key

JKTCBAGSMQIFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The enantioselective Heck arylation of 2,3-dihydrofuran with aryl iodides was studied.

Application

2,3-Dihydrofuran is a versatile reagent used in lanthanide-catalyzed Diels-Alder reactions with 2-pyrones and in Rh(II)-stabilized cycloadditions with vinylcarbenoids.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-11.2 °F - closed cup

Flash Point(C)

-24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia G Cobo-Angel et al.
Scientific reports, 9(1), 14025-14025 (2019-10-03)
Group B Streptococcus (GBS), is a leading cause of neonatal death and an emerging pathogen in adults. Additionally, GBS is a bovine pathogen causing intramammary infections. The likelihood of GBS interspecies transmission is largely unknown. We explored the potential transmission
The Journal of Organic Chemistry, 59, 4535-4535 (1994)
Lun-Zhi Dai et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(23), 7011-7018 (2008-07-08)
The gold(I)-catalyzed cycloisomerization of epoxy alkynes in the presence of a nucleophile is an efficient protocol to provide ketal skeletons with high stereoselectivity. An intramolecular reaction of propargylic/homopropargylic alcohols with oxirane to produce ketal/spiroketals in moderate yields under mild conditions
Raúl Pérez-Ruiz et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 5(1), 51-55 (2006-01-06)
The bicyclic oxetanes and resulting from photocycloaddition of aromatic aldehydes to 2,3-dihydrofuran, were efficiently cleaved by means of electron-transfer reduction, photoinduced by the electronically excited reductants 1-methoxynaphthalene (MN) and 2,7-dimethoxynaphthalene (DMN) in acetonitrile. The fluorescence quenching rates of DMN/MN by
Peizhong Xie et al.
Chemistry, an Asian journal, 7(7), 1533-1537 (2012-04-19)
A new domino reaction for Nazarov reagents: An efficient approach was developed for the construction of highly functionalized conjugated 2,3-dihydrofuran skeletons. Nazarov reagents were used for the first time in a phosphine-catalyzed domino reaction and successfully used to construct five-membered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service