Skip to Content
Merck
All Photos(1)

Key Documents

251615

Sigma-Aldrich

Ethylaluminum dichloride solution

1.0 M in hexanes

Synonym(s):

Dichloro(ethyl)alumane solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2AlCl2
CAS Number:
Molecular Weight:
126.95
Beilstein:
4123357
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

5 mmHg ( 60 °C)

form

liquid

concentration

1.0 M in hexanes

density

0.729 g/mL at 25 °C

SMILES string

CC[Al](Cl)Cl

InChI

1S/C2H5.Al.2ClH/c1-2;;;/h1H2,2H3;;2*1H/q;+2;;/p-2

InChI key

UAIZDWNSWGTKFZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Ethylaluminum dichloride is a strong Lewis acid and a proton scavenger that can be used:
  • To catalyze polymerization of isobutylene.
  • To facilitate Diels-Alder reaction of α,β-unsaturated esters.
  • To carry out Pinacol reduction rearrangements.
  • To promote Schmidt rearrangement of diketoazides in the synthesis of indolizidines and pyrroloazepinediones.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclorearrangement and cycloolefination of keto-bis-sulfones. A sulfone analog of a pinacol reduction-rearrangement.
Trost B M, et al.
Journal of the American Chemical Society, 114(13), 5432-5434 (1992)
Kinetic and Mechanistic Studies of the Polymerization of Isobutylene Catalyzed by EtAlCl2/Bis (2-chloroethyl) Ether Complex in Hexanes.
Banerjee S, et al.
Macromolecules, 48(16), 5474-5480 (2015)
Concise access to indolizidine and pyrroloazepine skeleta via intramolecular Schmidt reactions of azido 1, 3-diketones.
Lertpibulpanya D and Marsden S P
Organic & Biomolecular Chemistry, 4(18), 3498-3504 (2006)
Stereochemical aspects of the intramolecular Diels-Alder reactions of methyl deca-2, 7, 9-trienoates. 1. Thermal cyclizations.
Roush W R, et al.
The Journal of Organic Chemistry, 45(21), 4264-4267 (1980)
Ethylaluminum Dichloride.
Snider B B.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service