Skip to Content
Merck
All Photos(1)

Key Documents

256714

Sigma-Aldrich

Diethylaluminum chloride

97%

Synonym(s):

Chloro diethyl aluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2AlCl
CAS Number:
Molecular Weight:
120.56
Beilstein:
4123259
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

3 mmHg ( 60 °C)

Assay

97%

bp

125-126 °C/50 mmHg (lit.)

mp

−50 °C (lit.)

density

0.961 g/mL at 25 °C (lit.)

SMILES string

CC[Al](Cl)CC

InChI

1S/2C2H5.Al.ClH/c2*1-2;;/h2*1H2,2H3;;1H/q;;+1;/p-1

InChI key

YNLAOSYQHBDIKW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Diethylaluminum chloride is an organoaluminum reagent used in the reductive condensation of α-haloketones.

Application

Lewis-acid catalyst in Diels-Alder reactions and reactant in the synthesis of aluminum-phosphorus compounds.

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 49, 4073-4073 (1993)
Selective reactions with organoaluminum compounds
Hisashi Y et al.
Angewandte Chemie (International Edition in English), 17, 169-175 (1978)
Tetrahedron Letters, 35, 489-489 (1994)
Organometallics, 12, 3333-3333 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service