Skip to Content
Merck
All Photos(2)

Key Documents

250295

Sigma-Aldrich

1,8-Diiodooctane

98%, contains copper as stabilizer

Synonym(s):

Octamethylene diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I(CH2)8I
CAS Number:
Molecular Weight:
366.02
Beilstein:
1735437
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.5653 (lit.)

bp

167-169 °C/6 mmHg (lit.)

density

1.84 g/mL at 25 °C (lit.)

functional group

iodo

SMILES string

ICCCCCCCCI

InChI

1S/C8H16I2/c9-7-5-3-1-2-4-6-8-10/h1-8H2

InChI key

KZDTZHQLABJVLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,8-Diiodooctane has been employed as processing additive:
  • to improve the morphology and the efficiency of bulk heterojunctions solar cells, based on the regioregular poly(3-hexylthiophene) and a soluble fullerene derivative
  • to improve the power conversion efficiency of polymer solar cells

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiali Song et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(18), 2001986-2001986 (2020-10-02)
Morphological stability is crucially important for the long-term stability of polymer solar cells (PSCs). Many high-efficiency PSCs suffer from metastable morphology, resulting in severe device degradation. Here, a series of copolymers is developed by manipulating the content of chlorinated benzodithiophene-4,8-dione
Lichun Chang et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-10)
Semi-transparent organic solar cells (ST-OSCs) have attracted significant research attention, as they have strong potential to be applied in automobiles and buildings. For ST-OSCs, the transparent top electrode is an indispensable component, where the dielectric/metal/dielectric (D/M/D) structured electrode displayed a
Shuai Huang et al.
Journal of colloid and interface science, 583, 178-187 (2020-10-02)
A highly efficient inverted polymer solar cell (PSC) has been successfully demonstrated by utilizing a wide bandgap magnesium oxide (MgO) film and ZnO stacked structure as an effective cathode interfacial layer. The MgO/ZnO bilayer structure is designed to combine the
Soo Won Heo
Nanomaterials (Basel, Switzerland), 10(11) (2020-11-06)
In this study, we developed a method for fabricating ultrathin polymer substrates that can be used in ultra-flexible organic photovoltaics (OPVs) via a non-vacuum process using cyclic transparent optical polymer. In addition, a Ag nanowire network layer was used as
Wei Song et al.
Chemistry, an Asian journal, 14(9), 1472-1476 (2019-02-26)
Development of low-cost water-/alcohol-soluble interfacial materials is a crucial issue to promote the commercialization of polymer solar cells (PSCs). Herein, two derivatives of low-cost rhodamine, called sulforhodamine 101 (SR101) and sulforhodamine B (SRB), are applied as cathode interfacial layers (CILs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service