Skip to Content
Merck
All Photos(2)

Key Documents

142379

Sigma-Aldrich

4-tert-Butylpyridine

98%

Synonym(s):

4-(1,1-Dimethylethyl)pyridine, p-tert-Butylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N
CAS Number:
Molecular Weight:
135.21
Beilstein:
107594
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.495 (lit.)

bp

196-197 °C (lit.)

density

0.923 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1ccncc1

InChI

1S/C9H13N/c1-9(2,3)8-4-6-10-7-5-8/h4-7H,1-3H3

InChI key

YSHMQTRICHYLGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-tert-Butylpyridine is specific additive of redox electrolyte in dye sensitized solar cells and dye-sensitized TiO2 solar cells.

Application

4-tert-Butylpyridine was used in composition of electrolyte for dye-sensitized solar cell.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Firouzeh Ebadi et al.
Scientific reports, 9(1), 11962-11962 (2019-08-21)
In this research, we employed transient photo-voltage rise and decay measurements to investigate the origin of slow unsymmetrical rise and decay profiles in single and triple cation perovskite solar cells. Drastic changes in photo-voltage decay profile were observed upon insertion
Ishanie Rangeeka Perera et al.
Physical chemistry chemical physics : PCCP, 16(24), 12021-12028 (2014-01-08)
The abundance and low toxicity of manganese have led us to explore the application of manganese complexes as redox mediators for dye sensitized solar cells (DSCs), a promising solar energy conversion technology which mimics some of the key processes in
Fabrizio Giordano et al.
Nature communications, 7, 10379-10379 (2016-01-14)
Perovskite solar cells are one of the most promising photovoltaic technologies with their extraordinary progress in efficiency and the simple processes required to produce them. However, the frequent presence of a pronounced hysteresis in the current voltage characteristic of these
Firouzeh Ebadi et al.
Nature communications, 10(1), 1574-1574 (2019-04-07)
So-called negative capacitance seems to remain an obscure feature in the analysis of the frequency-dependent impedance of perovskite solar cells. It belongs to one of the puzzling peculiarities arising from the mixed ionic-electronic conductivity of this class of semiconductor. Here
Gerrit Boschloo et al.
The journal of physical chemistry. B, 110(26), 13144-13150 (2006-06-30)
Addition of 4-tert-butylpyridine (4TBP) to redox electrolytes used in dye-sensitized TiO2 solar cells has a large effect on their performance. In an electrolyte containing 0.7 M LiI and 0.05 M I2 in 3-methoxypropionitrile, addition of 0.5 M 4TBP gave an

Articles

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service