244465
1-Octyne
97%
Synonym(s):
1-Ethynylhexane, Hexylacetylene, n-Hexylacetylene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)5C≡CH
CAS Number:
Molecular Weight:
110.20
Beilstein:
1734494
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010411
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
37.7 mmHg ( 37.7 °C)
Assay
97%
form
liquid
impurities
≤3% 1-bromohexane
refractive index
n20/D 1.416 (lit.)
bp
127-128 °C (lit.)
mp
−80 °C (lit.)
density
0.747 g/mL at 25 °C (lit.)
SMILES string
CCCCCCC#C
InChI
1S/C8H14/c1-3-5-7-8-6-4-2/h1H,4-8H2,2H3
InChI key
UMIPWJGWASORKV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
1-Octyne was used as a mechanism-based inhibitor of AlkB (nonheme di-iron alkane monooxygenase).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
60.8 °F - closed cup
Flash Point(C)
16 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
D L Kline et al.
Medical and veterinary entomology, 21(4), 323-331 (2007-12-21)
Field studies were conducted at wooded wetlands in Gainesville, FL, U.S.A., to assess responses of natural populations of adult mosquitoes (Diptera: Culicidae) to American Biophysics MM-X and Coleman MD-2500 traps baited with enantiomers of 1-octen-3-ol, a naturally occurring compound, and
Joseph J Pesek et al.
Journal of chromatography. A, 992(1-2), 57-65 (2003-05-09)
The silanization/hydrosilation method is used to bond an alkyne (1-octyne) to a silica hydride surface. The new bonded material is characterized by elemental analysis and diffuse reflectance infrared Fourier transform spectroscopy. The hydrophobic behavior of this material was determined by
Alkyne-stabilized ruthenium nanoparticles: manipulation of intraparticle charge delocalization by nanoparticle charge States.
Xiongwu Kang et al.
Angewandte Chemie (International ed. in English), 49(49), 9496-9499 (2010-10-30)
Isaac S Marks et al.
Bioconjugate chemistry, 22(7), 1259-1263 (2011-05-05)
1,3-Dipolar [3 + 2] cycloaddition between azides and alkynes--an archetypal "click" chemistry--has been used increasingly for the functionalization of nucleic acids. Copper(I)-catalyzed 1,3-dipolar cycloaddition reactions between alkyne-tagged DNA molecules and azides work well, but they require optimization of multiple reagents
Jean-Charles Bruyere et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(34), 8061-8069 (2019-03-26)
The reactivity of ZnII dialkyl species ZnMe2 with a cyclic(alkyl)(amino)carbene, 1-[2,6-bis(1-methylethyl)phenyl]-3,3,5,5-tetramethyl-2-pyrrolidinylidene (CAAC, 1), was studied and extended to the preparation of robust CAAC-supported ZnII Lewis acidic organocations. CAAC adduct of ZnMe2 (2), formed from a 1:1 mixture of 1 and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service