Skip to Content
Merck
All Photos(3)

Key Documents

180211

Sigma-Aldrich

Borane tert-butylamine complex

powder, 97%

Synonym(s):

tert-Butylamine borane, NSC 114045, Trihydro(2-methyl-2-propanamine) boron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CNH2 · BH3
CAS Number:
Molecular Weight:
86.97
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

reagent type: reductant

mp

98-100 °C (dec.) (lit.)

functional group

amine

SMILES string

B.CC(C)(C)N

InChI

1S/C4H11N.BH3/c1-4(2,3)5;/h5H2,1-3H3;1H3

InChI key

GKFJEDWZQZKYHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Borane tert-butylamine complex is a mild reducing agent. It participates in the selective reduction of aldehydes and ketones to corresponding alcohol. It takes part as a reducing agent in the synthesis of monodisperse palladium nanoparticles (Pd NPs), monodisperse gold (Au) nanoparticles and carbon-supported Pd electrocatalysts.

Application

Borane tert-butylamine (BTB) complex may be used as strong co-reducing agent in the following syntheses:
  • copper (I) oxide nanoparticles
  • diphosphine-protected gold nanocluster
  • 4-acetoxycinnamyl alcohols
Reactant involved in:
  • Non-destructive analyses and conservation treatments of Indian drawings
  • Dehydrogenation reactions mediated by ruthenium bidentate phosphine complexes or ruthenium / iridium bis(N-heterocyclic carbene) complexes
  • The formation of σ-borane complexes
  • Ethanolysis of amine-borane adducts to form a reducing system for transfer hydrogenation of terminal olefins
  • Photochemical hydroboration and oxidation of single-walled carbon nanotubes

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Rae Kim et al.
Journal of nanoscience and nanotechnology, 13(9), 6027-6032 (2013-11-12)
To control the optical properties of Cu2O for a variety of application, we synthesized Cu2O in nanoscale without other treatments. Cu2O nanoparticles with an average size of 2.7 nm (sigma < or = 3.7%) were successfully synthesized in this study
John M Pettibone et al.
Physical chemistry chemical physics : PCCP, 14(12), 4142-4154 (2012-02-18)
We identify the reaction network governing gold monolayer protected cluster (MPC) formation during the reduction of Au(PPh(3))Cl and L(5) (L(5) = 1,5-bis(diphenylphosphino)pentane) in solutions. UV-vis spectroscopy and electrospray ionization mass spectrometry (ESI-MS) monitored the formation of ligated Au(x): 6 ≤
A facile synthesis of monodisperse Au nanoparticles and their catalysis of CO oxidation.
Peng S, et al.
Nano Research, 1(3), 229-234 (2008)
Facile synthesis of 4-hydroxycinnamyl p-coumarates.
Lu C and Raplph J.
Journal of Agricultural and Food Chemistry, 46(8), 2911-2913 (1998)
Reductive deoxygenation of aryl aldehydes and ketones by tert-butylamine-borane and aluminum chloride.
Lau CK, et al.
The Journal of Organic Chemistry, 54(2), 491-494 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service