Skip to Content
Merck
All Photos(3)

Documents

244066

Sigma-Aldrich

Nickel(II) acetate tetrahydrate

98%

Synonym(s):

Acetic acid nickel(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(OCOCH3)2 · 4H2O
CAS Number:
Molecular Weight:
248.84
Beilstein:
3730764
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

reaction suitability

core: nickel
reagent type: catalyst

density

1.798 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC(=O)O[Ni]OC(C)=O

InChI

1S/2C2H4O2.Ni.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

OINIXPNQKAZCRL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) acetate tetrahydrate is an ionic compound soluble in water and polar solvents. It is commonly used as a precursor for the synthesis of various nickel oxide and nickel hydroxide nanostructures. These nanostructures have applications in supercapacitors, batteries, and catalysis. Nickel(II) acetate tetrahydrate is also utilized in the production of nickel-based thin films and coatings through processes such as chemical vapor deposition and atomic layer deposition.

Application

Nickel(II) acetate tetrahydrate can be:    
  • Used as a precursor material in the hydrothermal synthesis of nanostructured nickel diselenide (NiSe2).     
  • Used as a precursor in the synthesis of the salamo-based dinuclear nickel(II) complex. 
  • Used as a precursor material in the synthesis of nickel oxide (NiO) via sol-gel method. Nickel oxide can be tailored for various applications, including electrochromic devices, gas sensors, and lithium-ion batteries.

Used in synthesis of polynuclear-nickel polyoxotungstate cluster compounds that are ideal for the design of molecular magnets.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Homa et al.
Scientific reports, 10(1), 740-740 (2020-01-22)
In this work, few-layered molybdenum disulfide (MoS2) was functionalized with metal oxide (MxOy) nanoparticles which served as a catalyst for carbon nanotubes (CNT) growth in the chemical vapour deposition (CVD) process. The resulting MoS2/MxOy/CNT functionalized nanomaterials were used for flame
Elham S Aazam et al.
Bioorganic chemistry, 57, 5-12 (2014-08-28)
Transition metal complexes compounds with Schiff bases ligand representing an important class of compounds that could be used to develop new metal-based anticancer agents and as precursors of metal NPs. Herein, 2,3-bis-[(3-ethoxy-2-hydroxybenzylidene)amino]but-2-enedinitrile Schiff base ligand and its corresponding copper/nickel complexes
K Korschelt et al.
Nanoscale, 10(27), 13074-13082 (2018-07-03)
The large-scale production and ecotoxicity of urea make its removal from wastewater a health and environmental challenge. Whereas the industrial removal of urea relies on hydrolysis at elevated temperatures and high pressure, nature solves the urea disposal problem with the
Clemente-Juan, J.M. et al.
Inorganic Chemistry, 38, 55-55 (1999)
Yong Jin Jeong et al.
ACS nano, 12(8), 7701-7709 (2018-07-20)
Photoresponsive transistor memories that can be erased using light-only bias are of significant interest owing to their convenient elimination of stored data for information delivery. Herein, we suggest a strategy to improve light-erasable organic transistor memories, which enables fast "photoinduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service