Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel(II) acetate tetrahydrate
Synonym(s):
Acetic acid nickel(II) salt
Linear Formula:
Ni(OCOCH
3
)
2
· 4H
2
O
CAS Number:
6018-89-9
Molecular Weight:
248.84
EC Number:
206-761-7
Products
Formula Weight
Physical Form
Purity
Reagent Type
Core
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
244066
Nickel(II) acetate tetrahydrate,
98%
Expand
View Pricing
379883
Nickel(II) acetate tetrahydrate,
99.995% trace metals basis
Expand
View Pricing
72225
Nickel(II) acetate tetrahydrate,
purum p.a., ≥99.0% (KT)
Expand
View Pricing