Skip to Content
Merck
All Photos(5)

Key Documents

151874

Sigma-Aldrich

Dimethyl sulfoxide-d6

Dimethyl sulfoxide-d6

99.9 atom % D

Synonym(s):

(Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein:
1237248
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.11
Pricing and availability is not currently available.

vapor pressure

0.42 mmHg ( 20 °C)

Quality Level

isotopic purity

99.9 atom % D

Assay

99% (CP)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %

technique(s)

NMR: suitable

impurities

≤0.0250% water
water

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Investigation of the Spatial Structure of Flufenamic Acid in Supercritical Carbon Dioxide Media via 2D NOESY.: This research explores the molecular interactions of flufenamic acid in supercritical CO2, highlighting analytical techniques using Dimethyl sulfoxide-d₆ as a solvent to enhance spectral readings (Khodov et al., 2023).

  • Taguchi Approach for Optimization of a Green Quantitative 1H-NMR Practice for Characterization of Levetiracetam and Brivaracetam in Pharmaceuticals.: Utilizes Dimethyl sulfoxide-d₆ in NMR spectroscopy to refine analytical methods for characterizing pharmaceuticals, aiming for environmental sustainability (Mansour et al., 2022).

  • Counterintuitive torsional barriers controlled by hydrogen bonding.: Investigates molecular torsion influenced by hydrogen bonding, with Dimethyl sulfoxide-d₆ employed to study solvent effects, contributing to our understanding of molecular dynamics in analytical chemistry (Barbero et al., 2020).

  • Elucidating Interactions between DMSO and Chelate-Based Ionic Liquids.: Examines the interaction dynamics between Dimethyl sulfoxide-d₆ and ionic liquids, offering insights into solvent-solute interactions critical in analytical methodologies (Chen et al., 2015).

  • Conformation of an octapeptide fragment (2-9) of kaliocin-1 in DMSO-d6 by 1H NMR and restrained molecular dynamics.: This study uses Dimethyl sulfoxide-d₆ in NMR to elucidate the conformational properties of a peptide, aiding in the understanding of peptide structure under analytical conditions (Sunilkumar et al., 2007).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Fortes et al.
Acta crystallographica Section B, Structural science, crystal engineering and materials, 76(Pt 5), 733-748 (2020-10-06)
Neutron powder diffraction data have been collected from a series of flash-frozen aqueous solutions of dimethyl sulfoxide (DMSO) with concentrations between 25 and 66.7 mol% DMSO. These reveal the existence of three stoichiometric hydrates, which crystallize on warming between 175 and
Mika Henrikki Sipponen et al.
Nature communications, 9(1), 2300-2300 (2018-06-14)
Dehydration reactions proceed readily in water-filled biological cells. Development of biocatalysts that mimic such compartmentalized reactions has been cumbersome due to the lack of low-cost nanomaterials and associated technologies. Here we show that cationic lignin nanospheres function as activating anchors
Satish N Dighe et al.
ACS omega, 5(48), 30971-30979 (2020-12-17)
Cholinesterases (ChE) are well-known drug targets for the treatment of Alzheimer's disease (AD). In continuation of work to develop novel cholinesterase inhibitors, we utilized a structure-based scaffold repurposing approach and discovered six novel ChE inhibitors from our recently developed DNA
Luís Novo et al.
Molecular pharmaceutics, 12(1), 150-161 (2014-11-11)
The applicability of small interfering RNA (siRNA) in future therapies depends on the availability of safe and efficient carrier systems. Ideally, siRNA delivery requires a system that is stable in the circulation but upon specific uptake into target cells can
Papawee Suabjakyong et al.
PloS one, 10(3), e0122733-e0122733 (2015-03-27)
The basidiomycetous mushroom Phellinus igniarius (L.) Quel. has been used as traditional medicine in various Asian countries for many years. Although many reports exist on its anti-oxidative and anti-inflammatory activities and therapeutic effects against various diseases, our current knowledge of

Articles

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Related Content

Widest range of NMR solvents with excellent chemical purity and the highest isotopic enrichment possible.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

See All

Questions

1–2 of 2 Questions  
  1. What are the differences between products 151874-50G-GL and 151874-50G-SB?

    1 answer
    1. The product or catalog number with "GL" at the end signifies a glass bottle with a screw cap. On the other hand, the product or catalog number with "SB" at the end represents a 50 ml serum bottle. Unlike the threaded glass bottle, the serum bottle features a silicon/PTFE septum which is secured by a 20 mm aluminum crimp seal without any tear feature.

      Helpful?

  2. AMP, GL, SB is what's diffrenent?

    1 answer
    1. AMP, GL, and SB all refer to the types of packaging for this product. The GL at the end of the product or catalog number indicates a glass bottle. This bottle has a screw cap. The AMP refers to ampoule packaging. The SB at the end of the product or catalog number indicates a 50 ml serum bottle. The bottle is not threaded. Rather there is a silicon/PTFE septum. The septum is held in place by a 20 mm aluminum crimp seal, no tear.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service