Skip to Content
Merck
All Photos(2)

Key Documents

235466

Sigma-Aldrich

1-Vinylimidazole

≥99%

Synonym(s):

1-Ethenyl-1H-imidazole, 1-Ethenylimidazole, 1-Vinyl-1H-imidazole, N-Vinylimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Beilstein:
105321
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

192-194 °C (lit.)
78-79 °C/13 mmHg (lit.)

density

1.039 g/mL at 25 °C (lit.)

SMILES string

C=Cn1ccnc1

InChI

1S/C5H6N2/c1-2-7-4-3-6-5-7/h2-5H,1H2

InChI key

OSSNTDFYBPYIEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Vinylimidazole has been employed:
  • as functional monomer in the synthesis of molecularly imprinted polymers[1]
  • as monomer in the preparation of magnetic-poly(divinylbenzene-1-vinylimidazole) [m-poly(DVB-VIM)] microbeads[2]
  • in the synthesis of novel ion imprinted polyvinylimidazole-silica hybrid copolymer (IIHC)[3]
  • in removal of the vinyl protecting group via isoprene-catalyzed lithiation process[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

183.2 °F - Pensky-Martens closed cup

Flash Point(C)

84 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N Pietruszka et al.
Biotechnology progress, 16(3), 408-415 (2000-06-03)
A new type of polymer, starch-modified by acrylamide, has been developed for application in aqueous two-phase systems (ATPS) for protein separation. Partial hydrolysis and acrylamide modification of starch to different degrees make it suitable for forming ATPS with poly(ethylene glycol)
Bilgen Osman et al.
Applied biochemistry and biotechnology, 168(2), 279-294 (2012-06-28)
Designing an immobilised metal ion affinity process on large-scale demands that a thorough understanding be developed regarding the adsorption behaviour of proteins on metal-loaded gels and the characteristic adsorption parameters to be evaluated. In view of this requirement, interaction of
Rackel Reis et al.
Scientific reports, 7(1), 4426-4426 (2017-07-02)
Thin-film composite poly(amide) (PA) membranes have greatly diversified water supplies and food products. However, users would benefit from a control of the electrostatic interactions between the liquid and the net surface charge interface in order to benefit wider application. The
Hong Wang et al.
Nature communications, 8, 13592-13592 (2017-01-05)
Nanoporous graphitic carbon membranes with defined chemical composition and pore architecture are novel nanomaterials that are actively pursued. Compared with easy-to-make porous carbon powders that dominate the porous carbon research and applications in energy generation/conversion and environmental remediation, porous carbon
Tetrahedron, 63, 947-947 (2007)

Questions

  1. Is this monomer stabilized by an inhibitor? If so, what is it?

    1 answer
    1. This product does not contain a stabilizer or inhibitor.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service