Skip to Content
Merck
All Photos(1)

Documents

D216402

Sigma-Aldrich

2,2′-Dipyridylamine

98%

Synonym(s):

bipyam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N3
CAS Number:
Molecular Weight:
171.20
Beilstein:
127131
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

222 °C/50 mmHg (lit.)

mp

90-92 °C (lit.)

SMILES string

N(c1ccccn1)c2ccccn2

InChI

1S/C10H9N3/c1-3-7-11-9(5-1)13-10-6-2-4-8-12-10/h1-8H,(H,11,12,13)

InChI key

HMMPCBAWTWYFLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2′-Dipyridylamine (bipyam) can be used:
  • As a bidentate N-donor ligand in the synthesis of various metal complexes.
  • To synthesize Pd-polyoxovanadates, heterogeneous catalyst for the oxidation of benzylic hydrocarbons.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of DNA, BSA, and HSA binding propensity of copper (II) complex with N-donor ligand 2, 2′-dipyridylamine
Kumar M and Masram DT
Polyhedron, 157, 511-520 (2019)
Mohan N Patel et al.
Journal of enzyme inhibition and medicinal chemistry, 26(2), 188-197 (2010-06-30)
The copper(II) complexes of the type [Cu(SPF)(A(n))Cl]/[Cu(PFL)(A(n))Cl] (where SPF is sparfloxacin, PFL is pefloxacin and A(n) is 2,2'-dipyridylamine/pyridine-2-carboxalehyde/thiophene-2-carboxaldehyde) were synthesised and were found to have a pyramidal geometry with a square base. The superoxide dismutase (SOD) like activity of the
Pierre Haquette et al.
Organic & biomolecular chemistry, 9(16), 5720-5727 (2011-06-23)
Organometallic complexes of the general formula [(η(6)-arene)Ru(N⁁N)Cl](+) and [(η(5)-Cp*)Rh(N⁁N)Cl](+) where N⁁N is a 2,2'-dipyridylamine (DPA) derivative carrying a thiol-targeted maleimide group, 2,2'-bispyridyl (bpy), 1,10-phenanthroline (phen) or ethylenediamine (en) and arene is benzene, 2-chloro-N-[2-(phenyl)ethyl]acetamide or p-cymene were identified as catalysts for
Development of copper (II)-diimine-iminodiacetate mixed ligand complexes as potential antitumor agents
Alvarez N, et al.
Inorgorganica Chimica Acta, 483, 61-70 (2018)
Mervat S Mohamed et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 86, 562-570 (2011-12-14)
In this study two new complexes [Cu(bpy)(Gly)Cl]·2H(2)O (1) and [Cu(dpa)(Gly)Cl]·2H(2)O (2) (bpy=2,2'-bipyridine; dpa=2,2'-dipyridylamine, Gly=glycine) have been synthesized and characterized by elemental analysis, IR, TGA, UV-vis and magnetic susceptibility measurements. The binding properties of the complexes with CT-DNA were investigated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service